Skip to Content
Merck
All Photos(1)

Documents

S6252

Supelco

Sulfapyridine

≥99.0%

Synonym(s):

4-Amino-N-[2-pyridyl]benzene sulfonamide, N1-(Pyridin-2-yl)sulfanilamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11N3O2S
CAS Number:
Molecular Weight:
249.29
Beilstein:
222065
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Quality Level

Assay

≥99.0%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Nc1ccc(cc1)S(=O)(=O)Nc2ccccn2

InChI

1S/C11H11N3O2S/c12-9-4-6-10(7-5-9)17(15,16)14-11-3-1-2-8-13-11/h1-8H,12H2,(H,13,14)

InChI key

GECHUMIMRBOMGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sulfapyridine is a metabolite of Sulfasalazine. It is an antibiotic and is mostly used in treating rheumatoid arthritis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sulfapyridine may be used as a reference standard in the determination of sulfapyridine in milk samples and plasma samples, using electrochemical magneto immunosensing assay technique and high performance liquid chromatography (HPLC), respectively.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of sulfapyridine and its major metabolites in plasma by high-pressure liquid chromatography.
Fischer C and Klotz U.
Journal of Chromatography. B, Biomedical Sciences and Applications, 146(1), 157-162 (1978)
Electrochemical magneto immunosensing of antibiotic residues in milk.
Zacco E, et al.
Biosensors And Bioelectronics, 22(9-10), 2184-2191 (2007)
Christopher Chidley et al.
Nature chemical biology, 7(6), 375-383 (2011-04-19)
We introduce an approach for detection of drug-protein interactions that combines a new yeast three-hybrid screening for identification of interactions with affinity chromatography for their unambiguous validation. We applied the methodology to the profiling of clinically approved drugs, resulting in
Cemalettin Alp et al.
Journal of enzyme inhibition and medicinal chemistry, 27(6), 818-824 (2011-12-23)
The inhibition of two human cytosolic carbonic anhydrase (hCA, EC 4.2.1.1) isozymes I, II and human serum isozyme VI, with a series of tosylited aromatic amine derivatives was investigated. The K(I) ranges of compounds 1-14 and acetazolamide against hCA I
Hai Haham et al.
Environmental science & technology, 46(21), 11870-11877 (2012-10-02)
Sorption-desorption behavior of sulfapyridine was studied with three distinct soil types low in organic carbon with or without the introduction of exogenous dissolved organic matter (DOM). Experiments with bulk soils yielded sorption coefficients equivalent to those obtained with soils richer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service