Skip to Content
Merck
All Photos(1)

Key Documents

S2180000

Sulindac

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(Z)-5-Fluoro-2-methyl-1-[p-(methylsulfinyl)benzylidene]indene-3-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H17FO3S
CAS Number:
Molecular Weight:
356.41
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sulindac

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1=C(CC(O)=O)c2cc(F)ccc2\C1=C/c3ccc(cc3)S(C)=O

InChI

1S/C20H17FO3S/c1-12-17(9-13-3-6-15(7-4-13)25(2)24)16-8-5-14(21)10-19(16)18(12)11-20(22)23/h3-10H,11H2,1-2H3,(H,22,23)/b17-9-

InChI key

MLKXDPUZXIRXEP-MFOYZWKCSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulindac EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Nonsteroidal anti-inflammatory; preferential inhibitor of COX-1.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The developmental toxicity of indomethacin and sulindac.
A Lione et al.
Reproductive toxicology (Elmsford, N.Y.), 9(1), 7-20 (1995-01-01)
Jigar Pravinchandra Modi et al.
Brain research, 1576, 91-99 (2014-06-27)
The present study analyzed whether administration of sulindac, a non-steroidal anti-inflammatory drug (NSAID) would prevent, attenuate or repair ischemia induced brain injury and reverse functional impairment in a focal ischemia model of stroke. Male Sprague-Dawley rats (weight 250-300 g) were
F M Giardiello
Cancer metastasis reviews, 13(3-4), 279-283 (1994-12-01)
Sulindac is useful in regression of adenomatous polyps. In addition to orally administered sulindac, rectal preparations also appear to be efficacious [32]. However, further studies are necessary to determine whether regression of adenomas, the precursor of colorectal cancer, will cause
D E Duggan
Drug metabolism reviews, 12(2), 325-337 (1981-01-01)
Sulindac is a prodrug which, following absorption, rapidly attains a metabolic equilibrium with its active pharmacophore, the sulfide metabolite. At the level of the whole body, the reversible interconversion sulindac in equilibrium sulfide, and the differing distributional and excretory properties
W R Waddell et al.
Journal of surgical oncology, 58(4), 252-256 (1995-04-01)
A putative explanation of the effect of sulindac on adenomatous colon and duodenal polyps from clinical observations and related in vitro experiments is presented. In cells with mutant APC genes, persistent high prostaglandin content of polyps leads to desensitization, downregulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service