Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C6H8N4O4
CAS Number:
Molecular Weight:
200.15
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
231-675-1
MDL number:
grade
analytical standard
Quality Segment
assay
≥95%
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
application(s)
forensics and toxicology
pharmaceutical (small molecule)
veterinary
format
neat
storage temp.
−20°C
SMILES string
Cn1c(COC(N)=O)ncc1N(=O)=O
InChI
1S/C6H8N4O4/c1-9-4(3-14-6(7)11)8-2-5(9)10(12)13/h2H,3H2,1H3,(H2,7,11)
InChI key
PQFRTXSWDXZRRS-UHFFFAOYSA-N
General description
Ronidazole is a nitroimidazole compound, commonly used as antiprotozoal and antibacterial agent.
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Ronidazole may be used as a reference standard in the determination of the analyte in poultry muscle and eggs using high-performance liquid chromatography with ultraviolet detection and confirmatory analysis by atmospheric pressure chemical ionization-mass spectrometry.
Storage Class
11 - Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis and antiprotozoal activity of 1-(3-chloro-2-hydroxypropyl)-substituted nitroimidazoles.
Hoffer Max and Grunberg E
Journal of Medicinal Chemistry, 17(9), 1019-1020 (1974)
Elizabeth J Kather et al.
Journal of veterinary internal medicine, 21(5), 966-970 (2007-10-18)
The nitroimidazole, ronidazole, has been demonstrated to have in vitro and in vivo activity against the protozoan Tritrichomonas foetus in cats. The purpose of this study was to evaluate the in vitro susceptibility of feline T. foetus isolates obtained from
I Burgener et al.
Schweizer Archiv fur Tierheilkunde, 151(8), 383-389 (2009-08-05)
Recent reports identified Tritrichomonas fetus, the causative agent of bovine trichomonosis, in cats with large-bowel diarrhea in the US. Between July 2007 and August 2008, a total of 105 Swiss cats were tested for T. fetus with the InPouchTM culture