Skip to Content
Merck

Skip To

PHR2012

Gabapentin Related Compound D

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

1-[(3-Oxo-2-azaspiro[4.5]dec-2-yl)methyl]cyclohexaneacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H29NO3
CAS Number:
Molecular Weight:
307.43
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

certified reference material, pharmaceutical secondary standard

agency

traceable to Ph. Eur. Y0001349, traceable to USP 1287358

API family

gabapentin

CofA

current certificate can be downloaded

packaging

pkg of 20 mg

application(s)

pharmaceutical

format

neat

storage temp.

2-30°C

SMILES string

OC(=O)CC1(CCCCC1)CN2CC3(CCCCC3)CC2=O

InChI

1S/C18H29NO3/c20-15-11-17(7-3-1-4-8-17)13-19(15)14-18(12-16(21)22)9-5-2-6-10-18/h1-14H2,(H,21,22)

InChI key

WJUZRUVUDOFJAK-UHFFFAOYSA-N

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP and EP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
To see an example of a Certificate of Analysis for this material enter LRAB8476 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
12873581287347PHR2011
application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

format

neat

format

neat

format

neat

format

-

Quality Level

300

Quality Level

-

Quality Level

-

Quality Level

300

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

storage temp.

2-30°C

storage temp.

-

storage temp.

-

storage temp.

-10 to -25°C

packaging

pkg of 20 mg

packaging

-

packaging

-

packaging

pkg of 100 mg


Still not finding the right product?

Explore all of our products under Gabapentin Related Compound D


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters