Skip to Content
Merck
All Photos(1)

Key Documents

PHR1760

Supelco

Nevirapine Related Compound A

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

11-Ethyl-5,11-dihydro-4-methyl-6H-dipyrido[3,2-b:2′,3′-e][1,4]diazepin-6-one, 5,11-DIHYDRO-6H-11-ETHYL- 4-METHYL-DIPYRIDO[3,2-b:2’,3’- e][1,4]DIAZEPIN-6-ONE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N4O
CAS Number:
Molecular Weight:
254.29
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1460725

API family

nevirapine

CofA

current certificate can be downloaded

packaging

pkg of 50 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

InChI

1S/C14H14N4O/c1-3-18-12-10(5-4-7-15-12)14(19)17-11-9(2)6-8-16-13(11)18/h4-8H,3H2,1-2H3,(H,17,19)

InChI key

HDVZWQWXAQRFKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Nevirapine Related Compound A is an impurity of nevirapine. Nevirapine is a non-nucleoside reverse transcriptase inhibitor (NNRTI), which belongs to the class of antiretroviral drugs. It acts by inhibiting the reverse transcriptase enzyme, thus stopping the multiplication of HIV. It is highly effective against advanced HIV disease.

Application

Nevirapine may be used as a pharmaceutical reference standard for the analysis of the analyte in pharmaceutical formulations and fixed dose combinations by chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA8553 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Sens. 1 - STOT RE 2 Inhalation

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of lamivudine, stavudine and nevirapine in antiretroviral fixed dose combinations by high performance liquid chromatography
Kapoor N, et al.
Analytica Chimica Acta, 570(1), 41-45 (2006)
MEKC determination of antiretroviral reverse transcriptase inhibitors lamivudine, stavudine, and nevirapine in pharmaceutical formulations
Sekar R and Azhaguvel S
Chromatographia, 67(5-6), 389-398 (2008)
Development and validation of a simple and rapid capillary zone electrophoresis method for determination of nnrti nevirapine in pharmaceutical formulations
Zanolli F, et al.
Journal of the Brazilian Chemical Society, 22(10), 2005-2012 (2011)
Nevirapine
USP42-NF37
United States Pharmacopeia/National Formulary, 32(3), 3101-3101 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service