Skip to Content
Merck
All Photos(1)

Key Documents

PHR1452

Supelco

Carprofen

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Carprofen, CAR, 6-Chloro-α-methyl-9H-carbazole-2-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12ClNO2
CAS Number:
Molecular Weight:
273.71
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000846
traceable to USP 1096699

API family

carprofen

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C(O)=O)c1ccc2c(c1)[nH]c3ccc(Cl)cc23

InChI

1S/C15H12ClNO2/c1-8(15(18)19)9-2-4-11-12-7-10(16)3-5-13(12)17-14(11)6-9/h2-8,17H,1H3,(H,18,19)

InChI key

PUXBGTOOZJQSKH-UHFFFAOYSA-N

Gene Information

human ... PTGS2(5743)

Looking for similar products? Visit Product Comparison Guide

General description

Carprofen is a nonsteroidal anti-inflammatory drug (NSAID), majorly used as equine medicine. It is known to be commonly present in the environment as a contaminant.
Pharmaceutical secondary standard for application in quality control. Provides pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Carprofen has been used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA3000 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel two-dimensional liquid chromatographic system for the online toxicity prediction of pharmaceuticals and related substances
Li L, et al.
Journal of Hazardous Materials, 293, 15-20 (2015)
Capillary electrophoresis for the analysis of non-steroidal anti-inflammatory drugs
Macia A, et al.
TrAC, Trends in Analytical Chemistry, 26(2), 133-153 (2007)
Nonsteroidal anti-inflammatory drug metabolism studies in horses in view of doping control: analytical strategies and challenges
Waraksa E, et al.
TrAC, Trends in Analytical Chemistry, 11(30), 3767-3792 (2019)
Danielle L Freeborn et al.
Toxicology and applied pharmacology, 282(2), 184-194 (2014-12-09)
The electroencephalogram (EEG) is an apical measure, capable of detecting changes in brain neuronal activity produced by internal or external stimuli. We assessed whether pesticides with different modes of action produced different changes in the EEG of adult male Long-Evans
K R Lezak et al.
Psychoneuroendocrinology, 45, 11-20 (2014-05-23)
Single nucleotide polymorphisms (SNP) in the genes for pituitary adenylate cyclase-activating polypeptide (PACAP) and the PAC1 receptor have been associated with several psychiatric disorders whose etiology has been associated with stressor exposure and/or dysregulation of the hypothalamic-pituitary-adrenal (HPA) axis. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service