Skip to Content
Merck
All Photos(1)

Key Documents

PHR1075

Supelco

Cimetidine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

SKF-92334

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N6S
CAS Number:
Molecular Weight:
252.34
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 475
traceable to Ph. Eur. C2175000
traceable to USP 1134062

API family

cimetidine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN\C(NC#N)=N\CCSCc1nc[nH]c1C

InChI

1S/C10H16N6S/c1-8-9(16-7-15-8)5-17-4-3-13-10(12-2)14-6-11/h7H,3-5H2,1-2H3,(H,15,16)(H2,12,13,14)

InChI key

AQIXAKUUQRKLND-UHFFFAOYSA-N

Gene Information

human ... HRH2(3274)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Cimetidine is an H2 receptor antagonist drug. It can inhibit the photosensitive response on the skin typically induced by most of the fluoroquinolones.

Application

Cimetidine may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7170 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Simultaneous determination of moxifloxacin and H2 receptor antagonist in pharmaceutical dosage formulations by RP-HPLC: application to in vitro drug interactions
Sultana N, et al.
Quimica nova, 34(4), 683-688 (2011)
Determination of cimetidine, famotidine, and ranitidine hydrochloride in the presence of their sulfoxide derivatives in pure and dosage forms by high-performance thin-layer chromatography and scanning densitometry
Kelani KM, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 85(5), 1015-1020 (2002)
P San Miguel et al.
Virchows Archiv : an international journal of pathology, 430(3), 261-263 (1997-03-01)
A case of carcinoma in the breast of a male patient with a chronic gastric ulcer is described. The patient had received cimetidine for 17 years. Histological examination of an excisional biopsy showed lobular in situ and infiltrating carcinoma. In
A Kumar
DICP : the annals of pharmacotherapy, 24(3), 289-295 (1990-03-01)
Suppressor T lymphocytes possess histamine2 (H2) receptors and contribute significantly to the function of the immune system. Experimentally, cimetidine, an H2-receptor antagonist, has been shown to enhance a variety of immunologic functions both in vivo and in vitro because of
A F Shinn
Drug safety, 7(4), 245-267 (1992-07-01)
The excellent efficacy and tolerability profiles of H2-antagonists have established these agents as the leading class of antiulcer drugs. Attention has been focused on drug interactions with H2-antagonists as a means of product differentiation and because many patients are receiving

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service