Skip to Content
Merck

Skip To

PHR1007

Mannitol

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

D-Mannitol, Mannite

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
HOCH2(CHOH)4CH2OH
CAS Number:
Molecular Weight:
182.17
NACRES:
NA.24
E Number:
E421
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
200-711-8
MDL number:
Beilstein/REAXYS Number:
1721898
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

certified reference material, pharmaceutical secondary standard

agency

traceable to Ph. Eur. M0200000, traceable to USP 1375105

API family

mannitol

CofA

current certificate can be downloaded

analyte chemical class(es)

oligosaccharides

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

mp

167-170 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-KVTDHHQDSA-N

General description

Mannitol is an osmotic diuretic with a variety of clinical applications, one of which is the reduction of intracranial pressure. It is also found to be protective against ischemic and nephrotoxic acute renal failure.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Mannitol may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using capillary isotachophoresis technique.

Biochem/physiol Actions

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Analysis Note

These secondary standards offer multi-traceability to the USP and EP (PhEur) primary standards, where they are available.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.
Ion Mobility: TWCCSN2 value of 132.3 Å2 [M-H]- The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer. For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com. Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage. TWCCS measurements are expected to be within 2% of this reference value. P/N PHR1007 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
To see an example of a Certificate of Analysis for this material enter LRAB7668 in the slot below. This is an example certificate only and may not be the lot that you receive.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
443907M190278513
description

Pharmaceutical Secondary Standard; Certified Reference Material

description

-

description

≥98% (GC), suitable for plant cell culture

description

≥99.9999% (metals basis), for boron determination

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

cell culture | plant: suitable

technique(s)

-

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

application(s)

-

application(s)

agriculture

application(s)

-

format

neat

format

-

format

-

format

-

Quality Level

300

Quality Level

100

Quality Level

200

Quality Level

100

grade

certified reference material, pharmaceutical secondary standard

grade

-

grade

-

grade

-

storage temp.

2-30°C

storage temp.

15-25°C

storage temp.

-

storage temp.

-


Still not finding the right product?

Explore all of our products under Mannitol


Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters