Skip to Content

P3200000

Prochlorperazine maleate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Prochlorperazine dimaleate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H24ClN3S · 2C4H4O4
CAS Number:
Molecular Weight:
606.09
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

prochlorperazine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)\C=C/C(O)=O.OC(=O)\C=C/C(O)=O.CN1CCN(CCCN2c3ccccc3Sc4ccc(Cl)cc24)CC1

InChI

1S/C20H24ClN3S.2C4H4O4/c1-22-11-13-23(14-12-22)9-4-10-24-17-5-2-3-6-19(17)25-20-8-7-16(21)15-18(20)24;2*5-3(6)1-2-4(7)8/h2-3,5-8,15H,4,9-14H2,1H3;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1-

InChI key

DSKIOWHQLUWFLG-SPIKMXEPSA-N

Gene Information

human ... DRD2(1813)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Prochlorperazine maleate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Phenothiazine antipsychotic; D2 dopamine receptor antagonist; antispasmodic

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Prochlorperazine maleate


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Lact. - Repr. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library