Skip to Content
Merck
All Photos(1)

Key Documents

P3143

Supelco

Poly(ethyleneimine) solution

analytical standard, 50 % (w/v) in H2O

Synonym(s):

Ethyleneimine polymer solution, PEI

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

mol wt

Mn ~60,000
Mw 750,000

concentration

50 % (w/v) in H2O

technique(s)

gel permeation chromatography (GPC): suitable

format

single component solution

SMILES string

N1CC1

InChI

1S/C2H5N/c1-2-3-1/h3H,1-2H2

InChI key

NOWKCMXCCJGMRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly(ethyleneimine) is a water soluble polymer used as a membrane lubricant for filtering frozen cell membranes. PEI cellulose in small columns will separate cyclic nucleotides from related compounds at the same electrolyte concentration that is used for thin layer chromatography on PEI cellulose-coated plates. PEI solution may also be used in the synthesis of a nanofibrous membrane, which may be used in the adsorption of anionic dyes and heavy metal ions from wastewater samples.[1]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ren-Hao Liu et al.
Molecular brain, 13(1), 41-41 (2020-03-18)
Sex differences in certain types of pain sensitivity and emotional responses have been previously reported. Synaptic plasticity is a key cellular mechanism for pain perception and emotional regulation, including long-term potentiation (LTP) and long-term depression (LTD). However, it is unclear
Eleonora Ciarlo et al.
Disease models & mechanisms, 6(2), 424-433 (2012-09-22)
Recent studies indicated that sortilin-related receptor 1 (SORL1) is a risk gene for late-onset Alzheimer's disease (AD), although its role in the aetiology and/or progression of this disorder is not fully understood. Here, we report the finding of a non-coding
Arseniy Gladkov et al.
Scientific reports, 7(1), 15625-15625 (2017-11-17)
The architecture of neuron connectivity in brain networks is one of the basic mechanisms by which to organize and sustain a particular function of the brain circuitry. There are areas of the brain composed of well-organized layers of neurons connected
Micro-nano structure poly (ether sulfones)/poly (ethyleneimine) nanofibrous affinity membranes for adsorption of anionic dyes and heavy metal ions in aqueous solution
Min M, et al.
Chemical Engineering Journal, 197, 88-100 (2012)
Kathrin S Grassme et al.
PloS one, 11(4), e0154176-e0154176 (2016-04-23)
Anterior gradient (AG) proteins have a thioredoxin fold and are targeted to the secretory pathway where they may act in the ER, as well as after secretion into the extracellular space. A newt member of the family (nAG) was previously

Questions

1–10 of 11 Questions  
  1. Is this PEI branched or linear?

    1 answer
    1. This product is an aqueous solution of a branched polyethyleneimine polymer.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. P3143, Poly(ethyleneimine) solution, analytical standard, 50 % (w/v) in H2O. Is gram to milliliter of  the w/v in this products, or other formulation,like mg to milliliter?

    1 answer
    1. This product is supplied as a 50% solution(w/v). The concentration is 50 grams PEI per 100 milliliters of H2O.

      To discuss this information further, please contact the Technical Service team. We kindly ask you to navigate to the link https://www.sigmaaldrich.com/techservice, click on "Product Technical Inquires" under the Products Section with all the required information so that a member of our team can reach out to you to assist further. Thank you.

      Helpful?

  5. What is the density of the procduct P3143 Supelco Poly(ethyleneimine) solution analytical standard, 50 % (w/v) in H2O ?

    1 answer
    1. According to the Safety Data Sheet, the density is 1.080 g/cm3.

      Helpful?

  6. There is no expiry date on the material bottle or CoA of P3143. Is there an expiry date for this product?

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment. For more information you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

  7. What is the molar concentration of Product P3143, Poly(ethyleneimine) solution?

    1 answer
    1. This product is nominally a 50% w/v solution of PEI in water, meaning 50 grams of poly(ethyleneimine) per 100 mL of solution. Concentrations of polymer solutions (such as PEI) are commonly reported either as a percentage or mg/mL, not in terms of molarity.

      Helpful?

  8. Do you have any references that describe using Product P3143, Poly(ethyleneimine) solution, in enzyme isolation or for transfections?

    1 answer
    1. For enzyme isolation, see:J.J. Jendrisak and R.R. Burgess, Biochem., 14(21), 4639-4645 (1975), "A new method for the large-scale purification of wheat germ DNA-dependent RNA polymerase II".This paper describes preparing a 10% stock solution, which was adjusted to pH 7.9 by adding hydrochloric acid.PEI has also been used for the precipitation of nucleic acids in the purification process of aminoacyl-tRNA synthetases:von der Haar, F., "Purification of Aminoacyl-tRNA Synthetases", Methods in Enzymology, 59, 257-267 (1979).For transfections, see the following references:Boussif, O. et al., "A versatile vector for gene and oligonucleotide transfer into cells in culture and in vivo: Polyethylenimine." Proc. Natl. Acad. Sci. USA, 92(16), 7297-7301 (1995).Thomas, M. and Klibanov, A.M., "Conjugation to gold nanoparticles enhances polyethylenimine's transfer of plasmid DNA into mammalian cells." Proc. Natl. Acad. Sci. USA, 100(16), 9138-9143 (2003).

      Helpful?

  9. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  10. What is the purity of Product P3143, Poly(ethyleneimine) solution?

    1 answer
    1. This product is a polymer with a large molecular weight distribution. It is not amenable to being analyzed by typical chromatographic assays such as HPLC or TLC. Therefore, we do not report an overall purity value for this product.

      Helpful?

1–10 of 11 Questions  

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service