Skip to Content
Merck
All Photos(1)

Key Documents

P2940000

Primaquine diphosphate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Primaquine bisphosphate, 8-(4-Amino-1-methylbutylamino)-6-methoxyquinoline diphosphate salt, Primaquine diphosphate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21N3O · 2H3PO4
CAS Number:
Molecular Weight:
455.34
Beilstein:
3812133
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

primaquine

manufacturer/tradename

EDQM

mp

205-206 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OP(O)(O)=O.OP(O)(O)=O.COc1cc(NC(C)CCCN)c2ncccc2c1

InChI

1S/C15H21N3O.2H3O4P/c1-11(5-3-7-16)18-14-10-13(19-2)9-12-6-4-8-17-15(12)14;2*1-5(2,3)4/h4,6,8-11,18H,3,5,7,16H2,1-2H3;2*(H3,1,2,3,4)

InChI key

GJOHLWZHWQUKAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Primaquine diphosphate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Muta. 2 - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael E von Fricken et al.
The American journal of tropical medicine and hygiene, 91(1), 77-80 (2014-04-30)
Administering primaquine (PQ) to treat malaria patients with glucose-6-phosphate dehydrogenase (G6PD) deficiency can pose a serious risk of drug-induced hemolysis (DIH). New easy to use point-of-care rapid diagnostic tests are being developed as an alternative to labor-intensive spectrophotometric methods, but
Salomón Durand et al.
The American journal of tropical medicine and hygiene, 91(1), 18-26 (2014-04-23)
We evaluated the efficacy of three primaquine (PQ) regimes to prevent relapses with Plasmodium vivax through an open-label randomized trial in Loreto, Peru. Vivax monoinfections were treated with chloroquine for 3 days and PQ in three different regimes: 0.5 mg/kg
Marli S Santana et al.
The American journal of tropical medicine and hygiene, 91(1), 74-76 (2014-05-29)
Glucose-6-phosphate dehydrogenase (G6PD) deficiency is one of the most common human genetic abnormalities, and it has a significant prevalence in the male population (X chromosome linked). The purpose of this study was to estimate the frequency of impaired fasting glucose
Low-dose primaquine for falciparum malaria.
Teun Bousema et al.
The Lancet. Infectious diseases, 14(8), 677-677 (2014-07-25)
Improving the radical cure of Plasmodium vivax malaria.
Ric N Price
The American journal of tropical medicine and hygiene, 91(1), 3-4 (2014-04-23)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service