Skip to Content
Merck
All Photos(1)

Key Documents

P1650000

Pilocarpine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(3S,4R)-4,5-Dihydro-3-ethyl-4-(1-methyl-1H-imidazol-5-ylmethyl)-2(3H)-furanone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16N2O2 · HCl
CAS Number:
Molecular Weight:
244.72
Beilstein:
4034491
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

pilocarpine

manufacturer/tradename

EDQM

mp

202-205 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CC[C@H]1[C@H](COC1=O)Cc2cncn2C

InChI

1S/C11H16N2O2.ClH/c1-3-10-8(6-15-11(10)14)4-9-5-12-7-13(9)2;/h5,7-8,10H,3-4,6H2,1-2H3;1H/t8-,10-;/m0./s1

InChI key

RNAICSBVACLLGM-GNAZCLTHSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pilocarpine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Nonselective muscarinic acetylcholine receptor agonist; used to produce an experimental model of epilepsy.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maxime Lévesque et al.
Seizure, 25, 18-25 (2015-02-04)
Mesial temporal lobe epilepsy (MTLE) is the most prevalent type of partial epileptic disorders. In this study, we have analyzed the impact of levetiracetam (LEV) in the pilocarpine model of MTLE. Sprague-Dawley rats (n=19) were injected with pilocarpine (380 mg/kg
Georgette L Suidan et al.
Arteriosclerosis, thrombosis, and vascular biology, 33(9), 2112-2120 (2013-07-05)
Aberrant blood-brain barrier (BBB) permeability is a hallmark pathology of many central nervous system diseases. von Willebrand factor (VWF) is stored in endothelial Weibel-Palade bodies from where it is released on activation into plasma and basement membrane. The role of
Rashmi M Risbud et al.
PloS one, 8(1), e53464-e53464 (2013-01-12)
MicroRNAs regulate protein synthesis by binding non-translated regions of mRNAs and suppressing translation and/or increasing mRNA degradation. MicroRNAs play an important role in the nervous system including controlling synaptic plasticity. Their expression is altered in disease states including stroke, head
Olav B Smeland et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 33(7), 1090-1097 (2013-04-25)
Although certain metabolic characteristics such as interictal glucose hypometabolism are well established for temporal lobe epilepsy (TLE), its pathogenesis still remains unclear. Here, we performed a comprehensive study of brain metabolism in a mouse model of TLE, induced by pilocarpine-status
Linda Holtman et al.
Epilepsia, 54(4), 589-595 (2013-02-13)
Brain inflammation occurs during epileptogenesis and may contribute to the development and progression of temporal lobe epilepsy. Recently, several studies have indicated that seizures may also increase specific blood plasma cytokine levels in animal models as well as in human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service