Skip to Content
Merck
All Photos(1)

Documents

P1300000

Phenytoin sodium

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5,5-Diphenylhydantoin sodium salt, 5,5-Diphenyl-2,4-imidazolidinedione, Phenytoin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C15H11N2O2Na
CAS Number:
Molecular Weight:
274.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

phenytoin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na]N1C(=O)NC(C1=O)(c2ccccc2)c3ccccc3

InChI

1S/C15H12N2O2.Na/c18-13-15(17-14(19)16-13,11-7-3-1-4-8-11)12-9-5-2-6-10-12;/h1-10H,(H2,16,17,18,19);/q;+1/p-1

InChI key

FJPYVLNWWICYDW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Phenytoin sodium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Reduces incidence of grand mal seizures; appears to stabilize excitable membranes perhaps through effects on Na+, K+, and Ca2+ channels.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shareef Ahmed et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 30(10), 823-830 (2014-08-29)
To determine whether penetrating scleral or corneal injury can enhance intraocular penetration of systemic moxifloxacin, vancomycin, and ceftazidime. Thirty rabbits were divided into 3 groups for each antibiotic and then further subdivided to receive either scleral or corneal injury to
Misty Good et al.
American journal of physiology. Gastrointestinal and liver physiology, 306(11), G1021-G1032 (2014-04-20)
Necrotizing enterocolitis (NEC) is the leading cause of death from gastrointestinal disease in premature infants and develops partly from an exaggerated intestinal epithelial immune response to indigenous microbes. There has been interest in administering probiotic bacteria to reduce NEC severity
Jan Hoffmann et al.
Neurobiology of disease, 74, 137-143 (2014-12-03)
To examine the effect of the orexinergic blockade with a dual orexin receptor antagonist (DORA) on experimental models of peripheral and central trigeminal as well as cortical activation relevant to migraine and migraine aura. In this study we used a
Maria B Rabaglino et al.
BMC genomics, 15, 1001-1001 (2014-11-21)
Major changes in gene expression occur in the fetal brain to modulate the function of this organ postnatally. Thus, factors can alter the genomics of the fetal brain, predisposing to neurological disorders later in life. We hypothesized that the physiological
Abhijeet Gummadavelli et al.
Epilepsia, 56(1), 114-124 (2014-12-03)
Understanding the neural mechanisms that support human consciousness is an important frontier in neuroscience and medicine. We previously developed a rodent model of temporal lobe seizures that recapitulates the human electroencephalography (EEG) signature of ictal and postictal neocortical slow waves

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service