Skip to Content
Merck
All Photos(3)

Key Documents

NIST17G

Sucrose Optical Rotation

NIST® SRM® 17g

Synonym(s):

Sucrose, α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
90825
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

packaging

bottle of 60 g

manufacturer/tradename

NIST® SRM®

17g

mp

185-187 °C (lit.)

application(s)

general analytical

format

neat

SMILES string

OC[C@H]1O[C@H](O[C@]2(CO)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12/h4-11,13-20H,1-3H2/t4-,5-,6-,7-,8+,9-,10+,11-,12+/m1/s1

InChI key

CZMRCDWAGMRECN-UGDNZRGBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SRM 17G is comprised of pure crystalline sucrose with certified value of chemical purity.

Each unit of SRM 17G includes a bottle containing ~60 g of sucrose.

SRM 17G_cert

SRM 17G_SDS

Application

Sucrose Optical Rotation standard reference material (SRM) is intended for use as a saccharimetry standard, used to calibrate polarimetric systems.

Features and Benefits

  • Accompanied with certificate documenting NIST-certified values of sucrose mass fraction and non-certified values of carbon and hydrogen mass fraction.
  • Certified chemical purity values are determined using a quantitative 1H-nuclear magnetic resonance spectroscopy (q1H-NMR).
  • Certified values are metrologically traceable to the International System of Units (SI) of mass.
  • The NIST certificate is provided with storage and use instructions.

Other Notes

Certified for the analytes listed below.
Sucrose

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service