Skip to Content
Merck
All Photos(1)

Key Documents

N2780

Sigma-Aldrich

1-Naphthol

BioXtra, ≥99%

Synonym(s):

α-Naphthol, 1-Hydroxynaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7OH
CAS Number:
Molecular Weight:
144.17
Beilstein:
1817321
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

4.5 (120 °C, vs air)

Quality Level

vapor pressure

1 mmHg ( 94 °C)

product line

BioXtra

Assay

≥99%

form

powder

autoignition temp.

1007 °F

expl. lim.

5 %

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

bp

278-280 °C (lit.)

mp

94-96 °C (lit.)

solubility

ethanol: 1 M, clear

anion traces

sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1cccc2ccccc12

InChI

1S/C10H8O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7,11H

InChI key

KJCVRFUGPWSIIH-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1A - STOT SE 2 Oral - STOT SE 3

Target Organs

Kidney, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anika Kasprick et al.
European journal of immunology, 45(5), 1462-1470 (2015-02-14)
The role of mast cells (MCs) in autoimmunity is the matter of an intensive scientific debate. Based on observations in different MC-deficient mouse strains, MCs are considered as fundamental players in autoimmune diseases. However, most recent data suggest that the
Hongbo Zhang et al.
Toxicology in vitro : an international journal published in association with BIBRA, 26(8), 1286-1293 (2012-01-24)
In vitro glucuronidation assays of diclofenac and indomethacin at pH 7.4 are biased by the instability of the glucuronides due to acyl migration. The extent of this acyl migration may be reduced significantly by performing the glucuronidation reaction at pH
Shitong Yang et al.
The Science of the total environment, 420, 214-221 (2012-02-15)
The increasing exploitation of multi-walled carbon nanotubes (MWCNTs) into many industrial processes has raised considerable concerns as they are likely to be released into the environment. The interactions of the pollutants in water with discharged MWNCTs will further influence the
Xiaoying Guo et al.
Environmental science & technology, 46(13), 7252-7259 (2012-06-09)
The sorption behavior of four hydrophobic organic contaminants (HOCs) (i.e., phenanthrene, naphthalene, lindane, and 1-naphthol) by three types of polymers namely polyethylene (PE), polystyrene (PS), and polyphenyleneoxide (PPO) was examined in this work. The organic carbon content-normalized sorption coefficients (K(oc))
Nenad Manevski et al.
Drug metabolism and disposition: the biological fate of chemicals, 40(11), 2192-2203 (2012-08-23)
The presence of bovine serum albumin (BSA) largely modulates the enzyme kinetics parameters of the human UDP-glucuronosyltransferase (UGT) 1A9, increasing both the apparent aglycone substrate affinity of the enzyme and its limiting reaction velocity (Drug Metab Dispos 39:2117-2129, 2011). For

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service