Skip to Content
Merck
All Photos(1)

Key Documents

N0700000

Nicotinic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3-Picolinic acid, Niacin, Pellagra preventive factor, Pyridine-3-carboxylic acid, Vitamin B3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
Beilstein:
109591
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

nicotinic acid

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

236-239 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)c1cccnc1

InChI

1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)

InChI key

PVNIIMVLHYAWGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nicotinic acid is a type of vitamin B that is effective against dyslipidemia. It can also be potentially used as a peripheral vasodilator.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nicotinic acid may be used as an EP reference standard for the determination of the analyte in pharmaceutical formulations by various chromatography techniques.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

379.4 °F - closed cup

Flash Point(C)

193 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous Estimation of Meclizine Hydrochloride and Nicotinic Acid in Pharmaceutical Dosage form by RP-HPLC Method
REDDY S, et al.
Asian Journal of Pharmaceutical Research and Health Care, 5(2) (2013)
Development and optimisation of a generic micellar electrokinetic capillary chromatography method to support analysis of a wide range of pharmaceuticals and excipients
Altria KD and McLean R
Journal of Pharmaceutical and Biomedical Analysis, 18(4-5), 807-813 (1998)
Nicotinic acid
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 5(2), 4961-4962 (2017)
RP-HPLC method for the determination of atorvastatin calcium and nicotinic acid in combined tablet dosage form
Shah DA, et al.
Indian Journal of Pharmaceutical Sciences, 69(5), 700-700 (2007)
Martina Lukasova et al.
Trends in pharmacological sciences, 32(12), 700-707 (2011-09-29)
Nicotinic acid (niacin) has been used for decades to prevent and treat atherosclerosis. The well-documented antiatherogenic activity is believed to result from its antidyslipidemic effects, which are accompanied by unwanted effects, especially a flush. There has been renewed interest in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service