Skip to Content
Merck
All Photos(1)

Key Documents

M1845000

Metrifonate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Trichlorfon, (2,2,2-Trichloro-1-hydroxyethyl)phosphonic acid dimethyl ester, Metrifonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8Cl3O4P
CAS Number:
Molecular Weight:
257.44
Beilstein:
1709434
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

trichlorfon, metrifonate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COP(=O)(OC)C(O)C(Cl)(Cl)Cl

InChI

1S/C4H8Cl3O4P/c1-10-12(9,11-2)3(8)4(5,6)7/h3,8H,1-2H3

InChI key

NFACJZMKEDPNKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Metrifonate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P N Tariot
The Journal of clinical psychiatry, 59 Suppl 9, 33-37 (1998-08-28)
Metrifonate, administered orally to patients with probable Alzheimer's disease in a once-daily dose, readily enters the brain and inhibits brain acetylcholinesterase (AChE) activity in a dose-dependent fashion. Metrifonate is a prodrug, converted non-enzymatically to 2,2-dichlorovinyl dimethyl phosphate, a long-acting inhibitor
H Feldmeier et al.
Acta tropica, 44(3), 357-368 (1987-09-01)
Metrifonate is an excellent drug for the treatment of urinary schistosomiasis in areas with S. haematobium monoinfection. Toxicity apparently is negligible. Side effects due to the inhibition of acetylcholinesterase are usually scarce, light and transient in nature. At the recommended
Metrifonate. A viewpoint.
E Giacobini
Drugs & aging, 11(6), 497-497 (1997-12-31)
B Holmstedt et al.
Archives of toxicology, 41(1), 3-29 (1978-10-13)
The organophosphorus compound 0,0-dimethyl-(1-hydroxy-2,2,2-trichloroethyl)-phosphonate was introduced as an insecticide, trichlorfon, in 1952 (Lorenz et al., 1955) and as a drug, metrifonate, in the treatment of schistosomiasis in 1960 (Lebrun and Cerf, 1960). This organophosphorus compound is unique in that it
B R Williams
American journal of health-system pharmacy : AJHP : official journal of the American Society of Health-System Pharmacists, 56(5), 427-432 (1999-03-30)
The pharmacology, pharmacokinetics, clinical efficacy, and adverse effects of metrifonate, a long-acting cholinesterase inhibitor, are discussed. Attempts to correct the central cholinergic deficits associated with Alzheimer's disease have included administration of cholinergic precursors, cholinergic agonists, and cholinesterase inhibitors. To date

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service