Skip to Content
Merck
All Photos(1)

Key Documents

M0120005

Malathion impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-[[Methoxy(methylthio)phosphinyl]thio]butanedioic acid 1,4-diethyl ester, Diethyl (2RS)-2-[(methoxy)(methylsulfanyl)-S-phosphinothioyl]butanedioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19O6PS2
CAS Number:
Molecular Weight:
330.36
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

malathion

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[P](=O)(SC(CC(=O)OCC)C(=O)OCC)(SC)OC

InChI

1S/C10H19O6PS2/c1-5-15-9(11)7-8(10(12)16-6-2)19-17(13,14-3)18-4/h8H,5-7H2,1-4H3

InChI key

LPQDGVLVYVULMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Malathion impurity A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

141.8 °F

Flash Point(C)

61 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franca M Buratti et al.
Journal of biochemical and molecular toxicology, 19(6), 406-414 (2006-01-20)
The organophosphorothioate (OPT) pesticide malathion (MAL) in mammals is readily hydrolyzed by mammalian carboxylesterases (CE). The reaction competes with the CYP-catalyzed formation of malaoxon (MOX), the toxic metabolite. Alterations or individual variations in CE activity may result in increased MOX
S Jianmongkol et al.
Toxicology and applied pharmacology, 139(2), 342-348 (1996-08-01)
The cholinergic toxicity of malathion is exacerbated by its isomerization product, isomalathion, which inhibits detoxifying carboxylesterases as well as target acetylcholinesterase (AChE). Previous work has shown that the four stereoisomers of isomalathion, (1R, 3R), (1R, 3S), (1S, 3R), and (1S
Jonathan A Doorn et al.
Chemical research in toxicology, 16(8), 958-965 (2003-08-20)
The present study was undertaken to test the hypothesis that acetylcholinesterase (AChE) inhibition by isomalathion stereoisomers proceeds with different primary leaving groups for (1R)- and (1S)-isomers. Consistent with results obtained with enzyme from other species, AChE from Torpedo californica (TcAChE)
S Xiong et al.
Journal of toxicology and environmental health, 51(2), 159-175 (1997-06-06)
In the present study, the effects of malathion and malathion derivatives on histamine and beta-hexosaminidase release by RBL-1 cells, rat peritoneal mast cells (RPMC), and human peripheral blood basophils (HPBB) and cutaneous mast calls were examined. One hour of incubation
J Błasiak et al.
Mutation research, 445(2), 275-283 (1999-11-27)
Malathion [S-(1,2-dicarboethoxyethyl)O,O-dimethyl phosphorodithioate] is a commonly used organophosphorus insecticide reported to be genotoxic both in vivo and in vitro, but the reports are conflicting. In order to elucidate the genotoxic potency of the main compounds present in commercial preparations of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service