Skip to Content
Merck

LAA10

L-Amino acids

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
85151701
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

analyte chemical class(es)

amino acids, peptides, proteins

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

10 Essential Amino Acids, 1 g of each:
  • A5131 L-Arginine Hydrochloride
  • H8125 L-Histidine Hydrochloride
  • I2752 L-Isoleucine
  • L8000 L-Leucine
  • L5626 L-Lysine Hydrochloride
  • M9625 L-Methionine
  • P2126 L-Phenylalanine
  • T8625 L-Threonine
  • T0254 L-Tryptophan
  • V0500 L-Valine


Still not finding the right product?

Explore all of our products under L-Amino acids


Storage Class

10 - Combustible liquids

wgk

nwg



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library