Skip to Content
Merck
All Photos(1)

Key Documents

IRMM651

Zinc 64Zn solution (certified for isotope abundance ratio)

IRMM®, certified reference material, 0.5 M in nitric acid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

certified reference material

Agency

IRMM®

isotopic purity

 64Zn

manufacturer/tradename

JRC

concentration

0.5 M in nitric acid

format

single component solution

mass shift

depleted

storage temp.

10-30°C

SMILES string

[64Zn]

InChI

1S/Zn/i1-1

InChI key

HCHKCACWOHOZIP-BJUDXGSMSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
IRMM651

Legal Information

IRMM is a registered trademark of European Commission

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service