Yes, the certified concenration of 1000 µg/mL is in accordance with Sulfide/S2.
Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
grade
analytical standard, certified reference material
CofA
current certificate can be downloaded
manufacturer/tradename
RTC ICS-023-100
concentration
1000 μg/mL
technique(s)
ion chromatography: suitable
application(s)
environmental
food and beverages
format
single component solution
storage temp.
2-8°C
General description
Analysis Note
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) ion chromatography: suitable | technique(s) ion chromatography: suitable | technique(s) ion chromatography: suitable | technique(s) ion chromatography: suitable |
| application(s) environmental | application(s) environmental | application(s) environmental | application(s) environmental |
| format single component solution | format single component solution | format single component solution | format single component solution |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 100, 300 |
| grade analytical standard, certified reference material | grade certified reference material, TraceCERT® | grade certified reference material, TraceCERT® | grade analytical standard |
| storage temp. 2-8°C | storage temp. - | storage temp. 10-30°C | storage temp. 2-30°C |
hcodes
pcodes
Hazard Classifications
Aquatic Chronic 2
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Is the 1000 ug/mL as per S2?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdfHelpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-
-
What is the packaging information for ICS023-500ML?
1 answer-
The packaging for product ICS023-500mL consists of a 500 ML NM 38-430 NAT HDPE BTL and a 38-430 RED PP CAP PTFE LINER.
Helpful?
-




