I0320301
Interferon α-2b
European Pharmacopoeia (EP) Reference Standard
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
interferon
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Interferon α-2b EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.
Packaging
The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.
Other Notes
Sales restrictions may apply.
related product
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Anterior segment OCT and confocal microscopy findings in atypical corneal intraepithelial neoplasia.
Cornea, 32(6), 875-879 (2013-03-30)
To report a case of biopsy-proven corneal intraepithelial neoplasia (CIN) diagnosed and followed clinically using high-resolution anterior segment optical coherence tomography (AS-OCT) and in vivo confocal microscopy (IVCM). Observational case report. A 57-year-old man presented with decreased vision in the
Journal of the American Academy of Dermatology, 68(1), 1-1 (2012-12-19)
The incidence of melanoma has increased for decades, and while surgical treatment of early stage disease is often curative, metastatic disease continues to carry a significantly less promising outlook with high associated health burden and economic cost. An expanding number
Zhonghua gan zang bing za zhi = Zhonghua ganzangbing zazhi = Chinese journal of hepatology, 20(8), 617-620 (2012-12-05)
To investigate a baculovirus insect cell system for expressing an interferon alpha 2b (IFNa2b)/immunoglobulin G-4 (IgG4) Fc fusion protein, which has long-acting antiviral effects. Human IFNa2b and IgG4 Fc cDNAs were generated by molecular cloning and inserted into a baculovirus
TSitologiia i genetika, 46(6), 12-18 (2013-01-05)
Spring rapeseed transgenic lines expressing human interferon alpha 2b were created by Agrobacterium-mediated transformation of aseptic plant leaf explants. The maximum antiviral activity of the leaf extracts reached 4500 IU/g fresh weight. It was determined that the antioxidant activity and
Zhurnal mikrobiologii, epidemiologii, i immunobiologii, (5)(5), 12-16 (2012-11-21)
Evaluate clinical features of the course of acute respiratory infection (ARI) and the state of microbiocenosis of nasopharynx of pregnant women at gestational term of 14 - 26 weeks. At the stage of outpatient therapy of ARI 49 pregnant women
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service