Skip to Content
Merck

Skip To

H2016

HPLC peptide standard mixture

analytical standard

Synonym(s):

Peptide calibration standard for HPLC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
85151701
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

description

Peptide standard

form

powder

composition

angiotensin II , Gly-Tyr , Leu enkephalin , Met enkephalin , Val-Tyr-Val

analyte chemical class(es)

amino acids, peptides, proteins

concentration

~0.5 mg each component (dried film)

technique(s)

HPLC: suitable

color

white

application(s)

food and beverages

compatibility

use to QA Reversed phase columns

format

multi-component solution

storage temp.

−20°C

Application

HPLC peptide standard mixture has been used for the separation of peptides of proteins in fish plasma samples using liquid chromatography coupled with quadrupole time of flight mass spectrometer (Q-TOF-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Amber vials containing a dried film composed of approx. 0.5 mg each of Gly-Tyr, Val-Tyr-Val, methionine enkephalin, leucine enkephalin and angiotensin II.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
H2899P811936978
format

multi-component solution

format

multi-component solution

format

multi-component solution

format

multi-component solution

form

powder

form

solid

form

liquid

form

-

technique(s)

HPLC: suitable

technique(s)

HPLC: suitable

technique(s)

gel permeation chromatography (GPC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

analyte chemical class(es)

amino acids, peptides, proteins

analyte chemical class(es)

amino acids, peptides, proteins

analyte chemical class(es)

amino acids, peptides, proteins

analyte chemical class(es)

-

concentration

~0.5 mg each component (dried film)

concentration

-

concentration

80 mg/mL (HSA and gamma-globulins)

concentration

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Product Information Sheet


Evelyn N Gitau et al.
Malaria journal, 10, 205-205 (2011-07-28)
A global proteomic strategy was used to identify proteins, which are differentially expressed in the murine model of severe malaria in the hope of facilitating future development of novel diagnostic, disease monitoring and treatment strategies. Mice (4-week-old CD1 male mice)
Lingyu Wang et al.
Electrophoresis, 41(21-22), 1832-1842 (2020-05-22)
Dynamic pH barrage junction focusing in CE enables effective signal enhancement, quantitative capture efficiencies, and straightforward optimization. The method is a technical variant of dynamic pH junction focusing. CE separation with dynamic pH barrage junction focusing is compatible with both
Plasma proteome profiles of White Sucker (Catostomus commersonii) from the Athabasca River within the oil sands deposit
Simmons.D.BD and Sherry.PJ
Comparative Biochemistry and Physiology. Part D, Genomics & Proteomics, 19, 181-189 (2016)



Global Trade Item Number

SKUGTIN
H2016-1VL04061835564132

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What animal are these peptides from?

    1 answer
    1. This product is synthetic and animal component-free. Please see the sample Certificate of Origin: https://www.sigmaaldrich.com/coo/undefined/H2016/*.

      Helpful?

Reviews

No rating value

Active Filters