Skip to Content
Merck
All Photos(1)

Documents

G0325000

Glybenclamide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-Chloro-N-[4-(cyclohexylureidosulfonyl)phenethyl]-2-methoxybenzamide, Glyburide, N-p-[2-(5-Chloro-2-methoxybenzamido)ethyl]benzenesulfonyl-N′-cyclohexylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H28ClN3O5S
CAS Number:
Molecular Weight:
494.00
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

glybenclamide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COc1ccc(Cl)cc1C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)NC3CCCCC3

InChI

1S/C23H28ClN3O5S/c1-32-21-12-9-17(24)15-20(21)22(28)25-14-13-16-7-10-19(11-8-16)33(30,31)27-23(29)26-18-5-3-2-4-6-18/h7-12,15,18H,2-6,13-14H2,1H3,(H,25,28)(H2,26,27,29)

InChI key

ZNNLBTZKUZBEKO-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Glybenclamide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nagaraj Kerur et al.
Investigative ophthalmology & visual science, 54(12), 7395-7401 (2013-10-12)
Accumulation of Alu RNA transcripts due to DICER1 deficiency in the retinal pigmented epithelium (RPE) promotes geographic atrophy. Recently we showed that Alu RNA activated the NLRP3 inflammasome, leading to RPE cell death via interleukin-18 (IL-18)-mediated MyD88 signaling. However, the
Kanako Sato et al.
Anesthesia and analgesia, 117(3), 627-633 (2013-07-23)
We previously reported the marked differences in the effects of sevoflurane anesthesia and propofol anesthesia on glucose use in fed rats; however, we could not elucidate mechanisms underlying the differences. We used fasted rats in this study. After surgical preparation
Kelly Barbosa Gama et al.
Journal of natural products, 76(4), 559-563 (2013-02-27)
Hecogenin is a sapogenin present in the leaves of species from the Agave genus, with a wide spectrum of reported pharmacological activities. The present study was undertaken to evaluate whether hecogenin acetate (1) has antinociceptive properties and to determine its
J P van Basten et al.
The Netherlands journal of medicine, 40(5-6), 305-307 (1992-06-01)
As sulphonylurea derivatives are used ubiquitously, the possibility that they may cause liver disease deserves attention. In contrast to the known pattern of reversible parenchymal liver disease due to sulphonylurea derivatives, we describe a fatal case of glyburide-induced cholestatic hepatitis
J F Caro
The American journal of medicine, 89(2A), 17S-25S (1990-08-20)
Increased hepatic glucose production is responsible for fasting hyperglycemia in type II diabetes. Insulin resistance is the key in this process because of the inability of insulin to suppress hepatic glucose production, thereby allowing an unopposed glucagon effect. Glyburide, one

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service