Skip to Content
Merck
All Photos(1)

Key Documents

F0030000

Felodipine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-(2,3-Dichlorophenyl)-1,4-dihydro-2,6-dimethyl-3,5-pyridinecarboxylic acid ethyl methyl ester, Plendil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19NO4Cl2
CAS Number:
Molecular Weight:
384.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

felodipine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)C1=C(C)NC(C)=C(C1c2cccc(Cl)c2Cl)C(=O)OC

InChI

1S/C18H19Cl2NO4/c1-5-25-18(23)14-10(3)21-9(2)13(17(22)24-4)15(14)11-7-6-8-12(19)16(11)20/h6-8,15,21H,5H2,1-4H3

InChI key

RZTAMFZIAATZDJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Felodipine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

L-type calcium channel blocker

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT RE 2 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W C Little et al.
Cardiovascular drugs and therapy, 9(5), 657-663 (1995-10-01)
Calcium entry through L-type calcium channels is essential for contraction of both arterial smooth muscle and the myocardium, and is important in cardiac conduction. First-generation calcium entry blockers lack or have a modest degree of vascular selectivity and inhibit cardiac
P A Todd et al.
Drugs, 44(2), 251-277 (1992-08-01)
Felodipine is a vascular-selective, dihydropyridine calcium antagonist previously investigated as a conventional tablet formulation administered twice daily. More recently considerable experience has been gained with an extended release (ER) formulation which has the convenience of once daily administration. Felodipine ER
Sayantan Chattoraj et al.
Journal of pharmaceutical sciences, 103(12), 3950-3957 (2014-10-30)
The nonisothermal crystallization kinetics of amorphous materials is routinely analyzed by statistically fitting the crystallization data to kinetic models. In this work, we systematically evaluate how the model-dependent crystallization kinetics is impacted by variations in the heating rate and the
A R Lorimer et al.
Journal of cardiovascular pharmacology, 15 Suppl 4, S85-S89 (1990-01-01)
Hypertensive patients, particularly the elderly, may often suffer from other diseases. Therefore, antihypertensive compounds should not negatively affect such disorders. Felodipine is a calcium antagonist that has potentially beneficial effects in angina pectoris and congestive heart failure. Further, it does
T O Morgan
Journal of cardiovascular pharmacology, 15 Suppl 4, S76-S84 (1990-01-01)
Felodipine is a dihydropyridine that blocks the slow entry channel for calcium. It is highly vascular selective and reduces blood pressure (BP) by dilatation of peripheral arterioles. It reduces BP in mild, moderate, and severe hypertension, and the fall in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service