Skip to Content
Merck

ERMBF424A

Maize GMO Standard ERM-BF424

ERM®, certified reference material, for the detection of GMO Maize (59122) with PCR

Synonym(s):

GMO Standard ERM-BF424

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

agency

ERM®

composition

maize (59122), 0%

manufacturer/tradename

JRC

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

matrix material

storage temp.

2-8°C

Analysis Note

For more information please see:
ERMBF424A

Legal Information

ERM is a registered trademark of European Commission


Still not finding the right product?

Explore all of our products under Maize GMO Standard ERM-BF424


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library