Skip to Content
Merck
All Photos(1)

Key Documents

E0110000

Enalapril for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Enalapril maleate salt, (S)-N-[1-(Ethoxycarbonyl)-3-phenylpropyl]-Ala-Pro maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28N2O5 · C4H4O4
CAS Number:
Molecular Weight:
492.52
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

enalapril

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H](C(N1[C@H](C(O)=O)CCC1)=O)N[C@H](CC(OCC)=O)CCC2=CC=CC=C2.O=C(OO)/C=C\C(O)=O

InChI

1S/C21H30N2O5.C4H4O5/c1-3-28-19(24)14-17(12-11-16-8-5-4-6-9-16)22-15(2)20(25)23-13-7-10-18(23)21(26)27;5-3(6)1-2-4(7)9-8/h4-6,8-9,15,17-18,22H,3,7,10-14H2,1-2H3,(H,26,27);1-2,8H,(H,5,6)/b;2-1-/t15-,17-,18-;/m0./s1

InChI key

UQKQUZOIQFOJOI-KJHLASMFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Enalapril for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

A long-acting angiotensin-converting enzyme inhibitor.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chrishan S Samuel et al.
Hypertension (Dallas, Tex. : 1979), 64(2), 315-322 (2014-05-29)
Relaxin is a naturally occurring peptide hormone that mediates systemic hemodynamic and renal adaptive changes during pregnancy and abrogates aberrant scar tissue formation (fibrosis) in diverse pathogeneses. However, its efficacy relative to renin–angiotensin system blockade, the most effective antifibrotic strategy
PARADIGM-HF--the experts' discussion.
Mariell Jessup et al.
The New England journal of medicine, 371(11), e15-e15 (2014-09-04)
Editorial comment.
Fernando Lorenzini
The Journal of urology, 192(6), 1883-1883 (2014-09-23)
Neprilysin inhibition for heart failure.
Alessio Galli et al.
The New England journal of medicine, 371(24), 2335-2335 (2014-12-11)
[The PARADIGM-HF study].
Luigi Tavazzi et al.
Giornale italiano di cardiologia (2006), 15(12), 651-655 (2014-12-24)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service