Skip to Content
Merck

DEN2003

Density Standard (20 °C)

UKAS ISO/IEC17025 and ISO 17034 certified, density: 0.7788 g/mL at 20 °C

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

agency

according to ASTM® D1480

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

density

0.7788 g/mL at 20 °C

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
industrial qc
personal care
petroleum

format

mixture

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon Density Standards can be found on the Paragon Scientific webpage

Application

This density CRM is for the calibration of instruments used to measure the density of materials that are fluids at the desired test temperature of 20 °C.

Features and Benefits

  • Manufactured and verified in an ISO 17025 and ISO 17034 accredited laboratory, as per UKAS
  • Carries an associated uncertainty of +/- 0.01%
  • Certified density value following ASTM D1480
  • Provided in a tamper-evident packaging to assure safe delivery
  • Marked holographically to ensure authenticity
  • Compliant to ASTM and IP test method protocols
  • Accompanied with a certificate of analysis (CoA) having information on expiry date and storage conditions for an effective audit

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials


Still not finding the right product?

Explore all of our products under Density Standard (20 °C)


signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

105.8 °F

flash_point_c

41 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library