Skip to Content

DELDATA

DEL Data for 1 analysis of DNA-Encoded Library information

Synonym(s):

DyNAbind® 10 Million Compound DNA-Encoded Library, DEL Hit Library, DEL Machine Learning

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352106
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


General description

Introducing our cutting-edge DNA Encoded Library (DEL) Extended Data Package, a revolutionary solution that brings unparalleled efficiency and innovation to the field of drug discovery. DEL Data combines the power of DNA tagging with advanced hit data analysis, enabling researchers to accelerate their drug development efforts and unlock new therapeutic possibilities. This DNA tagging allows for the parallel screening of millions or even billions of compounds, significantly enhancing the screening process′s speed and efficiency. The Extended Data Package takes this technology a step further by providing extensive and detailed information on the hits obtained from screening the DNA-encoded library. It offers researchers a comprehensive overview of the hit compounds′ chemical structures and binding affinities, empowering them to make informed decisions in selecting the most promising candidates for further investigation.

Application

DEL Data should be used in conjunction with DEL Dyna002

Legal Information

DyNAbind is a registered trademark of DyNAbind GmbH




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library