Skip to Content
Merck
All Photos(1)

Key Documents

D2955000

Domperidone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-(5-Chloro-2-oxo-1-benzimidazolinyl)-1-[3-(2-oxobenzimidazolinyl)propyl]piperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24ClN5O2
CAS Number:
Molecular Weight:
425.91
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

domperidone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Clc1ccc2N(C3CCN(CCCN4C(=O)Nc5ccccc45)CC3)C(=O)Nc2c1

InChI

1S/C22H24ClN5O2/c23-15-6-7-20-18(14-15)25-22(30)28(20)16-8-12-26(13-9-16)10-3-11-27-19-5-2-1-4-17(19)24-21(27)29/h1-2,4-7,14,16H,3,8-13H2,(H,24,29)(H,25,30)

InChI key

FGXWKSZFVQUSTL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Domperidone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A Prakash et al.
Drugs, 56(3), 429-445 (1998-10-20)
Domperidone is a selective antagonist at peripheral dopamine D2 receptors, with gastroprokinetic and antiemetic properties. It increases the frequency and duration of antral and duodenal contractions, thus decreasing/improving transit time of food through the gastrointestinal tract. Gastric emptying of liquids
Aravind Sugumar et al.
Clinical gastroenterology and hepatology : the official clinical practice journal of the American Gastroenterological Association, 6(7), 726-733 (2008-06-06)
Despite being widely used in more than 20 countries for the treatment of diabetic gastroparesis for several decades, domperidone is approved only on an investigational basis in the United States. However, because its use is increasing, it is important for
Savio C Reddymasu et al.
The American journal of gastroenterology, 102(9), 2036-2045 (2007-05-10)
Domperidone is a dopamine-2 receptor antagonist. It acts as an antiemetic and a prokinetic agent through its effects on the chemoreceptor trigger zone and motor function of the stomach and small intestine. Unlike metoclopramide, it does not cause any adverse
Domperidone.
M C Champion
General pharmacology, 19(4), 499-505 (1988-01-01)
Badriul Hegar et al.
The Turkish journal of pediatrics, 55(1), 1-7 (2013-05-23)
Gastroesophageal reflux (GER) is a normal physiologic process occurring several times per day in healthy infants. Regurgitation is defined as the passage of refluxed gastric contents into the pharynx or mouth, sometimes with expulsion out of the mouth. There are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service