Skip to Content
Merck
All Photos(1)

Key Documents

D0650000

Desmopressin

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H64N14O12S2
CAS Number:
Molecular Weight:
1069.22
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

desmopressin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

S1SCCC(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C(=O)NC(C1)C(=O)N4C(CCC4)C(=O)NC(CCCNC(=N)N)C(=O)NCC(=O)N)CC(=O)N)CCC(=O)N)Cc3ccccc3)Cc2ccc(cc2)O.OC(=O)C.O.O.O

InChI

1S/C46H64N14O12S2.C2H4O2.3H2O/c47-35(62)15-14-29-40(67)58-32(22-36(48)63)43(70)59-33(45(72)60-18-5-9-34(60)44(71)56-28(8-4-17-52-46(50)51)39(66)53-23-37(49)64)24-74-73-19-16-38(65)54-30(21-26-10-12-27(61)13-11-26)41(68)57-31(42(69)55-29)20-25-6-2-1-3-7-25;1-2(3)4;;;/h1-3,6-7,10-13,28-34,61H,4-5,8-9,14-24H2,(H2,47,62)(H2,48,63)(H2,49,64)(H,53,66)(H,54,65)(H,55,69)(H,56,71)(H,57,68)(H,58,67)(H,59,70)(H4,50,51,52);1H3,(H,3,4);3*1H2

InChI key

YNKFCNRZZPFMEX-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Desmopressin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

PLEASE NOTE: TEMPORARILY UNAVAILABLE DUE TO RESTRICTED QUANTITY

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathan T Connell et al.
Blood advances, 5(1), 301-325 (2021-02-12)
von Willebrand disease (VWD) is a common inherited bleeding disorder. Significant variability exists in management options offered to patients. These evidence-based guidelines from the American Society of Hematology (ASH), the International Society on Thrombosis and Haemostasis (ISTH), the National Hemophilia
Patrina Hy Caldwell et al.
The Cochrane database of systematic reviews, 5, CD002911-CD002911 (2020-05-05)
Enuresis (bedwetting) affects up to 20% of five-year-olds and can have considerable social, emotional and psychological effects. Treatments include alarms (activated by urination), behavioural interventions and drugs. To assess the effects of enuresis alarms for treating enuresis in children. We
D A Vassiliadi et al.
The Journal of clinical endocrinology and metabolism, 101(12), 4878-4885 (2016-09-24)
Cushing's disease (CD) has a significant relapse rate after successful transsphenoidal surgery (TSS). Many CD patients respond aberrantly to the desmopressin test (DT). Disappearance of this response after surgery may suggest complete removal of abnormal corticotrophs and a lower possibility
Marco Ranucci et al.
Journal of cardiothoracic and vascular anesthesia, 31(5), 1588-1594 (2017-08-06)
Hemodilution has been associated with both hypocoagulability and hypercoagulability in studies based on thromboelastography (TEG). Severe hemodilution during cardiopulmonary bypass (CPB) is a risk factor for morbidity in cardiac surgery. This study investigated the effects of different degrees of hemodilution
Naoya Egami et al.
Laboratory investigation; a journal of technical methods and pathology, 93(9), 1001-1011 (2013-07-24)
The purpose of this study was to clarify the underlying mechanism of vertiginous attacks in Ménière's disease (MD) while obtaining insight into water homeostasis in the inner ear using a new animal model. We conducted both histopathological and functional assessment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service