Skip to Content
Merck

Skip To

COND75

Conductivity Standard - 75 μmhos/cm

certified reference material

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

CofA

current certificate can be downloaded

manufacturer/tradename

RTC COND-75-0.500

concentration

in H2O

application(s)

environmental
food and beverages
pharma/biopharma processes
water monitoring

format

single component solution

storage temp.

2-30°C

General description

The product is a certified reference material (CRM), produced and certified in accordance with ISO 17034 and ISO/IEC 17025 requirements. The assigned specific conductance value of 75.0 ± 0.2 µmhos/cm is traceable to SI and higher order standards from NIST through an unbroken chain of comparisons.

All information regarding the use of this CRM can be found on the certificate of analysis (CoA).

This ready-to-use sample is delivered in 500 mL HDPE bottle.

Application

Conductivity calibration standard solution to qualify conductivity measurement devices.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
COND25COND147COND50
application(s)

environmental
food and beverages
pharma/biopharma processes
water monitoring

application(s)

environmental
food and beverages
pharmaceutical
water monitoring

application(s)

environmental
food and beverages
pharma/biopharma processes
water monitoring

application(s)

environmental

format

single component solution

format

single component solution

format

single component solution

format

single component solution

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

Quality Level

100, 300

Quality Level

300

Quality Level

300

Quality Level

300


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters