Skip to Content
Merck

COND600

Conductivity Standard - 600 μmhos/cm

certified reference material

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

CofA

current certificate can be downloaded

manufacturer/tradename

RTC COND-600-0.500

concentration

in H2O

application(s)

environmental

format

single component solution

storage temp.

2-30°C

General description

This certified reference material (CRM) is designed, manufactured and validated for accuracy and stability following ISO 17034 and ISO/IEC 17025. The specific conductance value of 600.0 ± 3 µmhos/ cm is quantified using SI and higher order standards from NIST through an unbroken chain of comparisons.

The certificate of analysis (CoA) includes information such as traceability, uncertainty, stability, packaging & handling, and safety.

Thisready-to-use sample is delivered in 500 mL HDPE bottle.

Application

Conductivity standard solution for calibration checks of conductivity measurement equipment.

Reported for use as a standard in the development of custom electrical conductivity sensors for chemical mapping of ammonia release.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library