Skip to Content
Merck
All Photos(1)

Key Documents

CCW4103

Millipore

Malt Extract Acidified Broth pH 4,2

Liquid medium for cultivation of yeasts and molds for MF methods, suitable for microbiology

Synonym(s):

MEA (pH 4.3)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.74

Quality Level

sterility

sterile

form

liquid

shelf life

9 mo. at 8‑25 °C

feature

ready-to-use

composition

malt extract, 17.0 g/L

packaging

box of 50 tubes

storage condition

(Tightly closed)

parameter

2 mL filling volume

capacity

2 mL

tube volume

3 mL

color

yellowish

pH

4.3±0.1 (25 °C)

application(s)

food and beverages
microbiology

suitability

molds
yeasts

General description

Malt Extract Acidified Broth (pH 4.3) is a modified version of the classic Malt Extract Broth according to Reiss, where the pH has been lowered to obtain greater selectivity of the medium.

The pH should not be lowered below 3.5 as these conditions are difficult and delay growth a lot. The culture medium should also not overheat.

Application

Malt Extract Acidified Broth is suitable for the cultivation of yeasts and molds for MF methods.

Other Notes

The material and quality of the membrane filter affect the microorganism’s recovery significantly.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service