Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12171500
NACRES:
NA.47
Product Name
5(6)-Carboxyfluorescein diacetate Mixed isomers, ≥90% (HPLC)
Quality Segment
assay
≥90% (HPLC)
form
powder
color
white to yellow
solubility
acetone: 49.00-51.00 mg/mL, clear, colorless to yellow
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
−20°C
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.