Skip to Content
Merck
All Photos(1)

Key Documents

C2580000

Colchicine

(European Pharmacopoeia (EP) Reference Standard)

Synonym(s):

(S)-N-(5,6,7,9-Tetrahydro-1,2,3,10-tetramethoxy-9-oxobenzo[a]heptalen-7-yl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H25NO6
CAS Number:
Molecular Weight:
399.44
Beilstein:
2228813
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

colchicine

manufacturer/tradename

EDQM

mp

150-160 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COC1=CC=C2C(=CC1=O)[C@H](CCc3cc(OC)c(OC)c(OC)c23)NC(C)=O

InChI

1S/C22H25NO6/c1-12(24)23-16-8-6-13-10-19(27-3)21(28-4)22(29-5)20(13)14-7-9-18(26-2)17(25)11-15(14)16/h7,9-11,16H,6,8H2,1-5H3,(H,23,24)/t16-/m0/s1

InChI key

IAKHMKGGTNLKSZ-INIZCTEOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Colchicine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Muta. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Lu et al.
Pharmaceutical research, 29(11), 2943-2971 (2012-07-21)
Tubulin dynamics is a promising target for new chemotherapeutic agents. The colchicine binding site is one of the most important pockets for potential tubulin polymerization destabilizers. Colchicine binding site inhibitors (CBSI) exert their biological effects by inhibiting tubulin assembly and
Alberto Massarotti et al.
ChemMedChem, 7(1), 33-42 (2011-10-13)
Computational approaches have been increasingly applied to drug design over the past three decades and have already provided some useful results in the discovery of anticancer drugs. Given the increased availability of crystal structures in recent years, a growing number
Massimo Imazio et al.
Heart failure reviews, 18(3), 355-360 (2012-06-05)
Empiric anti-inflammatory therapy for acute and recurrent pericarditis is warranted for viral and idiopathic cases that represent most cases of pericarditis in developed countries. For specific uncomplicated etiologies, such as systemic autoimmune diseases and postpericardiotomy syndromes, the same drugs are
I Grattagliano et al.
Clinical pharmacology and therapeutics, 95(1), 89-97 (2013-07-23)
Familial Mediterranean fever (FMF), an inherited autosomal recessive disorder, is characterized by sporadic, paroxysmal attacks of fever and serosal inflammation, lasting 1-3 days. Patients may develop renal amyloidosis, arthritis, serositis, and skin and oral lesions. Diagnosis is based on clinical
Massimo Imazio et al.
Heart (British Cardiac Society), 98(14), 1078-1082 (2012-03-24)
The purpose of this study was to investigate the efficacy and safety of colchicine for pericarditis prevention. Recurrent pericarditis prevention is a major management goal that may reduce morbidity and management costs. Although empiric anti-inflammatory therapy is considered the mainstay

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service