Skip to Content
Merck
All Photos(1)

Key Documents

C2269000

Clindamycin phosphate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Clindamycin 2-phosphate, Clindamycin 2-dihydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34ClN2O8PS
CAS Number:
Molecular Weight:
504.96
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clindamycin

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCC[C@@H]1C[C@H](N(C)C1)C(=O)NC([C@H](C)Cl)C2O[C@H](SC)[C@H](OP(O)(O)=O)[C@@H](O)[C@H]2O

InChI

1S/C18H34ClN2O8PS/c1-5-6-10-7-11(21(3)8-10)17(24)20-12(9(2)19)15-13(22)14(23)16(18(28-15)31-4)29-30(25,26)27/h9-16,18,22-23H,5-8H2,1-4H3,(H,20,24)(H2,25,26,27)/t9-,10+,11-,12?,13+,14-,15?,16+,18+/m0/s1

InChI key

UFUVLHLTWXBHGZ-MWBQRTRKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clindamycin phosphate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Spectrum of Activity: Gram positive cocci and taxoplasma. Especially active against anaerobic bacteria.
Mode of Action: Inhibits protein synthesis in bacterial by binding the 50s ribosomal subunit.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Antibacterial and antiprotozoal antibiotic of the licosamide class.
Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Lact. - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shao-Min Wang et al.
Rapid communications in mass spectrometry : RCM, 23(6), 899-906 (2009-02-18)
A simple high-performance liquid chromatography/electrospray ionization tandem mass spectrometric (HPLC/ESI-MS/MS) method has been developed for the rapid identification of clindamycin phosphate and its degradation products or related impurities in clindamycin phosphate injection. Detection was performed by quadrupole time-of-flight mass spectrometry
C Hascicek et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 77(1), 116-121 (2010-11-23)
The aim of this work was to study the clindamycin release kinetics from floating delivery systems consisting of two modules assembled in void configuration, according to the modified release technology platform known as Dome Matrix®. Two modules differently shaped, i.e.
Michael T Jarratt et al.
Journal of drugs in dermatology : JDD, 11(3), 318-326 (2012-03-08)
Topical combination therapy containing a retinoid and an antimicrobial is an effective treatment for acne vulgaris. To evaluate the efficacy and safety of a new topical formulation containing clindamycin phosphate 1.2% and tretinoin 0.025% solubilized in an aqueous-based gel (CT
L Babayeva et al.
Journal of the European Academy of Dermatology and Venereology : JEADV, 25(3), 328-333 (2010-07-30)
No single effective topical treatment is available for treating all pathogenic factors causing acne vulgaris (AV). Salicylic acid (SA), tretinoin (all-TRA) and clindamycin phosphate (CDP) are known to to be effective agents depending on their comedolytic and anti-inflammatory properties. To
Asuka Munehiro et al.
The Journal of dermatological treatment, 23(3), 172-176 (2011-07-16)
Cosmetic moisturisers were applied to one side of the face of 18 male Japanese patients with acne vulgaris who were treated with a topical administration of adapalene and clindamycin phosphate gels. We assessed the alleviating effect of the moisturisers on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service