Skip to Content
Merck
All Photos(1)

Key Documents

C0410000

Canrenone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(17α)-17-Hydroxy-3-oxopregna-4,6-diene-21-carboxylic acid γ-lactone, NSC 261713

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28O3
CAS Number:
Molecular Weight:
340.46
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

canrenone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C[C@@]12C(C=C[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]45OC(CC5)=O)=CC(CC1)=O

InChI

1S/C22H28O3/c1-20-9-5-15(23)13-14(20)3-4-16-17(20)6-10-21(2)18(16)7-11-22(21)12-8-19(24)25-22/h3-4,13,16-18H,5-12H2,1-2H3/t16-,17+,18+,20+,21+,22-/m1/s1

InChI key

UJVLDDZCTMKXJK-WNHSNXHDSA-N

Gene Information

human ... NR3C2(4306)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Canrenone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiang Tian et al.
Hypertension (Dallas, Tex. : 1979), 54(6), 1313-1320 (2009-11-04)
Spironolactone has been noted to attenuate cardiac fibrosis. We have observed that the cardiotonic steroid marinobufagenin plays an important role in the diastolic dysfunction and cardiac fibrosis seen with experimental renal failure. We performed the following studies to determine whether
Alessandro Boccanelli et al.
Journal of cardiovascular medicine (Hagerstown, Md.), 8(9), 683-691 (2007-08-19)
Excess aldosterone activity contributes to the pathogenesis and progression of heart failure (HF). Aldosterone antagonists improve clinical outcome in patients with severe HF or left ventricular (LV) dysfunction after myocardial infarction, but knowledge of their impact in mild chronic HF
Maysa Faisal Suyagh et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 878(9-10), 769-776 (2010-02-16)
A selective and sensitive liquid chromatography (LC)-atmospheric pressure chemical ionisation (APCI)-mass spectroscopic (MS) assay of canrenone has been developed and validated employing Dried Blood Spots (DBS) as the sample collection medium. DBS samples were prepared by applying 30 microl of
Da-Ming Huang et al.
Journal of biomedicine & biotechnology, 2011, 917232-917232 (2011-12-29)
A procedure for simultaneous identification and quantification of canrenone and its biotransformed product 11-α-hydroxy-canrenone by high-performance liquid chromatography with ultraviolet detector (HPLC-UVD) and mass spectrometry (LC-MS) methods was proposed. The optimal determination variables on the HPLC-UVD or LC-MS coupled with
Haijuan Dong et al.
Journal of mass spectrometry : JMS, 41(4), 477-486 (2006-03-17)
A sensitive and specific liquid chromatography-atmospheric pressure chemical ionization-mass spectrometry (LC-APCI-MS) method for the simultaneous determination of spironolactone and its active metabolite canrenone in human plasma has been developed and validated. After the addition of estazolam as the internal standard

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service