Skip to Content
Merck

Skip To

BP901

Phenobarbital

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

5-Ethyl-5-phenyl-2,4,6-pyrimidinetrione, 5-Ethyl-5-phenylbarbituric acid, Luminal

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C12H12N2O3
CAS Number:
Molecular Weight:
232.24
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
233363
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

phenobarbital

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

drug control

kontrollierte Droge in Deutschland, psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal); Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

CCC1(C(=O)NC(=O)NC1=O)c2ccccc2

InChI

1S/C12H12N2O3/c1-2-12(8-6-4-3-5-7-8)9(15)13-11(17)14-10(12)16/h3-7H,2H2,1H3,(H2,13,14,15,16,17)

InChI key

DDBREPKUVSBGFI-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Phenobarbital BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Phenobarbital Injection
  • Phenobarbital Elixir
  • Phenobarbital Tablets
  • Paediatric Phenobarbital Oral Solution
  • Biochem/physiol Actions

    Anesthetic; sedative; hypnotic; anticonvulsant

    Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Legal Information

    German
    Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

    English
    This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.

    Compare Similar Items

    View Full Comparison

    Show Differences

    1 of 1

    This Item
    PHR8843P1636Y0001351
    grade

    pharmaceutical primary standard

    grade

    certified reference material, pharmaceutical secondary standard

    grade

    -

    grade

    pharmaceutical primary standard

    manufacturer/tradename

    BP

    manufacturer/tradename

    -

    manufacturer/tradename

    -

    manufacturer/tradename

    EDQM

    application(s)

    pharmaceutical
    pharmaceutical small molecule

    application(s)

    pharmaceutical small molecule

    application(s)

    -

    application(s)

    pharmaceutical (small molecule)

    form

    solid

    form

    -

    form

    powder

    form

    -

    format

    neat

    format

    -

    format

    -

    format

    neat

    shelf life

    limited shelf life, expiry date on the label

    shelf life

    -

    shelf life

    -

    shelf life

    -


    Still not finding the right product?

    Explore all of our products under Phenobarbital


    pictograms

    Skull and crossbonesHealth hazard

    signalword

    Danger

    Hazard Classifications

    Acute Tox. 3 Oral - Carc. 2 - Repr. 1B - Skin Sens. 1

    Storage Class

    6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

    wgk

    WGK 3

    flash_point_f

    Not applicable

    flash_point_c

    Not applicable



    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library



    Questions

    Reviews

    No rating value

    Active Filters