Skip to Content
Merck

BP803

Zidovudine

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

3′-Azido-3′-deoxythymidine, AZT, Azidothymidine, ZDV, Zidovudine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H13N5O4
CAS Number:
Molecular Weight:
267.24
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
3595791
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

zidovudine

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

113-115 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

CC1=CN([C@H]2C[C@H](N=[N+]=[N-])[C@@H](CO)O2)C(=O)NC1=O

InChI

1S/C10H13N5O4/c1-5-3-15(10(18)12-9(5)17)8-2-6(13-14-11)7(4-16)19-8/h3,6-8,16H,2,4H2,1H3,(H,12,17,18)/t6-,7+,8+/m0/s1

InChI key

HBOMLICNUCNMMY-XLPZGREQSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Zidovudine BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Abacavir, Zidovudine and Lamivudine Tablets
  • Zidovudine and Lamivudine Tablets
  • Zidovudine Capsules
  • Zidovudine Infusion
  • Abacavir Tablets
  • Zidovudine Tablets
  • Biochem/physiol Actions

    Reverse transcriptase inhibitor active against HIV-1 virus.

    Packaging

    Unit quantity: 150 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.


    Still not finding the right product?

    Explore all of our products under Zidovudine


    pictograms

    Health hazard

    signalword

    Warning

    hcodes

    Hazard Classifications

    Carc. 2 - Muta. 2

    Storage Class

    11 - Combustible Solids

    wgk

    WGK 1

    flash_point_f

    Not applicable

    flash_point_c

    Not applicable



    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library