Skip to Content
Merck

Skip To

BP802

Lamivudine

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

2′,3′-Dideoxy-3′-thiacytidine, 3TC, Epivir, Epivir-HBV, Heptovir, Zeffix

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C8H11N3O3S
CAS Number:
Molecular Weight:
229.26
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

lamivudine

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

NC1=NC(=O)N(C=C1)[C@@H]2CS[C@H](CO)O2

InChI

1S/C8H11N3O3S/c9-5-1-2-11(8(13)10-5)6-4-15-7(3-12)14-6/h1-2,6-7,12H,3-4H2,(H2,9,10,13)/t6-,7+/m0/s1

InChI key

JTEGQNOMFQHVDC-NKWVEPMBSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lamivudine BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Abacavir, Zidovudine and Lamivudine Tablets
  • Zidovudine and Lamivudine Tablets
  • Abacavir and Lamivudine Tablets
  • Lamivudine Tablets
  • Biochem/physiol Actions

    Lamivudine is a potent nucleoside analog reverse transcriptase inhibitor (nRTI).
    Lamivudine is a potent nucleoside analog reverse transcriptase inhibitor (nRTI). It is an analogue of cytidine, and can inhibit both types (1 and 2) of HIV reverse transcriptase as well as the reverse transcriptase of hepatitis B. It needs to be phosphorylated to its triphosphate form before it is active. 3TC-triphosphate also inhibits cellular DNA polymerase.

    Packaging

    Unit quantity: 200 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Compare Similar Items

    View Full Comparison

    Show Differences

    1 of 1

    This Item
    Y00004251356836Y0000518
    Lamivudine British Pharmacopoeia (BP) Reference Standard

    BP802

    Lamivudine

    manufacturer/tradename

    BP

    manufacturer/tradename

    EDQM

    manufacturer/tradename

    USP

    manufacturer/tradename

    EDQM

    grade

    pharmaceutical primary standard

    grade

    pharmaceutical primary standard

    grade

    pharmaceutical primary standard

    grade

    pharmaceutical primary standard

    application(s)

    pharmaceutical
    pharmaceutical small molecule

    application(s)

    pharmaceutical (small molecule)

    application(s)

    pharmaceutical (small molecule)

    application(s)

    pharmaceutical (small molecule)

    form

    solid

    form

    -

    form

    -

    form

    -

    shelf life

    limited shelf life, expiry date on the label

    shelf life

    -

    shelf life

    -

    shelf life

    -

    format

    neat

    format

    neat

    format

    neat

    format

    neat


    Still not finding the right product?

    Explore all of our products under Lamivudine


    pictograms

    Health hazard

    signalword

    Warning

    Storage Class

    11 - Combustible Solids

    flash_point_f

    Not applicable

    flash_point_c

    Not applicable

    target_organs

    Blood

    Hazard Classifications

    Aquatic Chronic 3 - Repr. 2 - STOT RE 2



    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library



    Questions

    Reviews

    No rating value

    Active Filters