Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C8H15N7O2S3
CAS Number:
Molecular Weight:
337.45
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
grade
pharmaceutical primary standard
agency
BP
API family
famotidine
form
solid
shelf life
limited shelf life, expiry date on the label
packaging
pkg of 100 mg
manufacturer/tradename
BP
storage condition
OK to freeze
color
white
mp
162-165 °C
solubility
water: insoluble
application(s)
pharmaceutical
pharmaceutical small molecule
format
neat
shipped in
ambient
storage temp.
2-8°C
SMILES string
N\C(N)=N\c1nc(CSCCC(=N)NS(N)(=O)=O)cs1
InChI
1S/C8H15N7O2S3/c9-6(15-20(12,16)17)1-2-18-3-5-4-19-8(13-5)14-7(10)11/h4H,1-3H2,(H2,9,15)(H2,12,16,17)(H4,10,11,13,14)
InChI key
XUFQPHANEAPEMJ-UHFFFAOYSA-N
Gene Information
human ... HRH2(3274)
General description
Famotidine is a hydrophilic, cationic, histamine H2 receptor antagonist drug that effectively inhibits gastric acid secretion in humans.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Famotidine BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.
Also used in monographs such as:Famotidine Tablets
Also used in monographs such as:
Biochem/physiol Actions
H2 histamine receptor antagonist; anti-ulcer agent
Packaging
Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia
Other Notes
Sales restrictions may apply.
Still not finding the right product?
Explore all of our products under Famotidine
Storage Class
11 - Combustible Solids
WGK
WGK 2
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.