Skip to Content
Merck

BP413

Pseudoephedrine hydrochloride

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

(+)-Pseudoephedrine hydrochloride, (+)-ψ-Ephedrine hydrochloride, (1S,2S)-2-Methylamino-1-phenylpropanol hydrochloride, d-Isoephedrine hydrochloride, d-Pseudoephedrine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5CH[CH(NHCH3)CH3]OH·HCl
CAS Number:
Molecular Weight:
201.69
EC Number:
206-462-1
UNSPSC Code:
12352210
NACRES:
NA.24
Beilstein/REAXYS Number:
3915112
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

pseudoephedrine

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

185-188 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

Cl.CN[C@@H](C)[C@@H](O)c1ccccc1

InChI

1S/C10H15NO.ClH/c1-8(11-2)10(12)9-6-4-3-5-7-9;/h3-8,10-12H,1-2H3;1H/t8-,10+;/m0./s1

InChI key

BALXUFOVQVENIU-KXNXZCPBSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pseudoephedrine hydrochloride BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Pseudoephedrine Tablets
  • Pseudoephedrine Oral Solution
  • Biochem/physiol Actions

    Non-selective adrenergic agonist; decongestant

    Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.


    Still not finding the right product?

    Explore all of our products under Pseudoephedrine hydrochloride


    pictograms

    Exclamation mark

    signalword

    Warning

    hcodes

    Hazard Classifications

    Acute Tox. 4 Oral - STOT SE 3

    target_organs

    Central nervous system

    Storage Class

    11 - Combustible Solids

    wgk

    WGK 3

    flash_point_f

    Not applicable

    flash_point_c

    Not applicable



    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library