Skip to Content
Merck

BP205

Isoprenaline hydrochloride

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

(±)-Isoproterenol hydrochloride, 1-(3′,4′-Dihydroxyphenyl)-2-isopropylaminoethanol hydrochloride, N-Isopropyl-DL-noradrenaline hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H17NO3 · HCl
CAS Number:
Molecular Weight:
247.72
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
3917363
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

isoprenaline

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

165-175 °C (dec.) (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

Cl.CC(C)NCC(O)c1ccc(O)c(O)c1

InChI

1S/C11H17NO3.ClH/c1-7(2)12-6-11(15)8-3-4-9(13)10(14)5-8;/h3-5,7,11-15H,6H2,1-2H3;1H

InChI key

IROWCYIEJAOFOW-UHFFFAOYSA-N

Gene Information

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isoprenaline hydrochloride BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Isoprenaline Injection
  • Biochem/physiol Actions

    β-adrenoceptor agonist; increases cytosolic cAMP.

    Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.


    Still not finding the right product?

    Explore all of our products under Isoprenaline hydrochloride


    Storage Class

    11 - Combustible Solids

    WGK

    WGK 2

    Flash Point (°F)

    Not applicable

    Flash Point (°C)

    Not applicable



    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library