Skip to Content
Merck

BP003

Adrenaline acid tartrate

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

(−)-Epinephrine (+)-bitartrate salt, L-3,4-Dihydroxy-α-(methylaminomethyl)benzyl alcohol D-hydrogen bitartrate salt, L-Adrenaline (+)-bitartrate salt, L-Adrenaline D-hydrogentartrate, L-Epinephrine D-hydrogentartrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H13NO3 · C4H6O6
CAS Number:
Molecular Weight:
333.29
UNSPSC Code:
12352106
NACRES:
NA.24
EC Number:
200-097-1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

adrenaline

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O.CNC[C@H](O)c1ccc(O)c(O)c1

InChI

1S/C9H13NO3.C4H6O6/c1-10-5-9(13)6-2-3-7(11)8(12)4-6;5-1(3(7)8)2(6)4(9)10/h2-4,9-13H,5H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t9-;1-,2-/m01/s1

InChI key

YLXIPWWIOISBDD-NDAAPVSOSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Adrenaline acid tartrate BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Bupivacaine and Adrenaline Injection / Bupivacaine and Epinephrine Injection
  • Lidocaine and Adrenaline Injection / Lidocaine and Epinephrine Injection
  • Adrenaline Injection/Epinephrine Injection
  • Dilute Adrenaline Injection 1 in 10,000/Dilute Epinephrine Injection 1 in 10,000
  • Adrenaline and Cocaine Intranasal Solution / Epinephrine and Cocaine Intranasal Solution
  • Noradrenaline Injection / Norepinephrine Injection
  • Adrenaline Solution / Epinephrine Solution
  • Adrenaline Eye Drops / Epinephrine Eye Drops
  • Biochem/physiol Actions

    Adrenergic receptor agonist.

    Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.


    pictograms

    Skull and crossbones

    signalword

    Danger

    hcodes

    Hazard Classifications

    Acute Tox. 1 Oral

    Storage Class

    6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

    wgk

    WGK 3

    flash_point_f

    Not applicable

    flash_point_c

    Not applicable



    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library