Skip to Content
Merck
All Photos(1)

Key Documents

BCR266

7H-Dibenzo[c,g]carbazole

BCR®, certified reference material

Synonym(s):

3,4,5,6-Dibenzocarbazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H13N
CAS Number:
Molecular Weight:
267.32
Beilstein:
213015
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-8°C

SMILES string

[H]n1c2ccc3ccccc3c2c4c1ccc5ccccc45

InChI

1S/C20H13N/c1-3-7-15-13(5-1)9-11-17-19(15)20-16-8-4-2-6-14(16)10-12-18(20)21-17/h1-12,21H

InChI key

STJXCDGCXVZHDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR266

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zuzana Valovicová et al.
Mutation research, 665(1-2), 51-60 (2009-05-12)
Liver progenitor (oval) cells are a potential target cell population for hepatocarcinogens. Our recent study showed that the liver carcinogens 7H-dibenzo[c,g]carbazole (DBC) and 5,9-dimethyldibenzo[c,g]carbazole (DiMeDBC), but not the sarcomagen N-methyldibenzo[c,g]carbazole (N-MeDBC), induced several cellular events associated with tumor promotion in
G Ericson et al.
Mutation research, 454(1-2), 11-20 (2000-10-18)
The time-course and dose dependent formation of DNA adducts in juvenile northern pike (Esox lucius) following a single exposure to a mixture of benzo[a]pyrene (BaP), benzo[k]fluoranthene (BkF) and 7H-dibenzo[c,g]carbazole (DBC) were investigated by use of the (32)P-postlabelling assay. A complex
Glenn Talaska et al.
Toxicology letters, 162(2-3), 246-249 (2005-11-01)
Studies of the impact of phase 1 enzyme polymorphisms on genetic damage have yielded mixed results. We studied how genetic damage would be altered when specific genes were ablated under low dose conditions. Knockouts (KO) were generated from c57bl6/J mice
Jan Vondrácek et al.
Mutation research, 596(1-2), 43-56 (2006-01-13)
Immature liver progenitor cells have been suggested to be an important target of hepatotoxins and hepatocarcinogens. The goal of the present study was to assess the impact of 7H-dibenzo[c,g]carbazole (DBC) and its tissue-specific carcinogenic N-methyl (N-MeDBC) and 5,9-dimethyl (DiMeDBC) derivatives
D L Gray et al.
Experimental lung research, 27(3), 245-253 (2001-04-11)
Polycyclic aromatic hydrocarbons (PAH) and N-heterocyclic aromatic hydrocarbons (NHA) are environmental pollutants formed during the incomplete combustion of organic materials. Benzo(a)pyrene (BaP) and 7H-dibenzo(c,g)carbazole (DBC) are well-characterized representatives of the PAH and NHA classes of compunds, respectively. Both are demonstrated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service