Skip to Content
Merck
All Photos(1)

Key Documents

B0950000

Betadex

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

β-Cyclodextrin, Caraway, Cycloheptaamylose, Cyclomaltoheptaose, Schardinger β-Dextrin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H70O35
CAS Number:
Molecular Weight:
1134.98
Beilstein:
78623
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

betadex

optical activity

[α]20/D +162±3°, c = 1.5% in H2O

manufacturer/tradename

EDQM

mp

290-300 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]8CO)O[C@H]1[C@H](O)[C@H]2O

InChI

1S/C42H70O35/c43-1-8-29-15(50)22(57)36(64-8)72-30-9(2-44)66-38(24(59)17(30)52)74-32-11(4-46)68-40(26(61)19(32)54)76-34-13(6-48)70-42(28(63)21(34)56)77-35-14(7-49)69-41(27(62)20(35)55)75-33-12(5-47)67-39(25(60)18(33)53)73-31-10(3-45)65-37(71-29)23(58)16(31)51/h8-63H,1-7H2/t8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-/m1/s1

InChI key

WHGYBXFWUBPSRW-FOUAGVGXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Betadex EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Golfo G Kordopati et al.
Biochimica et biophysica acta, 1850(1), 159-168 (2014-12-03)
Cyclodextrins (CDs) in combination with therapeutic proteins and other bioactive compounds have been proposed as candidates that show enhanced chemical and enzymatic stability, better absorption, slower plasma clearance and improved dose-response curves or immunogenicity. As a result, an important number
Omair Aleem et al.
Journal of pharmaceutical and biomedical analysis, 47(3), 535-540 (2008-03-28)
The solid-state properties, dissolution profile and antimicrobial activity of inclusion complexes of cefdinir (CEF) with beta-cyclodextrin (betaCD) and hydroxypropyl beta-cyclodextrin (HPbetaCD) were investigated. The phase solubility profiles of cefdinir with betaCD and HPbetaCD were classified as A(L)-type, which indicates the
H Chen et al.
Se pu = Chinese journal of chromatography, 17(6), 533-538 (2003-01-30)
A review of beta-cyclodextrin bonded stationary phases in chiral liquid chromatography is given, focusing on design and synthesis of beta-cyclodextrin bonded chiral stationary phases. Fifty-three references are presented in this paper.
Nuclear magnetic resonance methods for high molecular weight proteins: a study involving a complex of maltose binding protein and beta-cyclodextrin.
L E Kay
Methods in enzymology, 339, 174-203 (2001-07-21)
Yuanqi Lu et al.
Pakistan journal of pharmaceutical sciences, 27(3), 487-490 (2014-05-09)
A method was proposed to determine kaempferol and quercetin in Hippophae rhamnoides L medicinal preparation xindakang tablet by β-cyclodextrin modified micellar electrokinetic capillary chromatography. Under the optimized conditions: buffer solution of 20 mmol/L Na(2)B(4)O(7)-KH(2)PO(4) (pH 9.0)-20mmol/L SDS-6mmol/L β-CD-5%(v/v) MeCN, applied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service