Skip to Content
Merck

Skip To

A6877

Agarose

Medium EEO, Molecular Biology

Synonym(s):

3,6-Anhydro-α-L-galacto-β-D-galactan, Agarose ME

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
41105317
PubChem Substance ID:
EC Number:
232-731-8
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

algae (marine)

Quality Segment

grade

Molecular Biology

type

Type II

form

powder

impurities

≤10% moisture content

EEO

0.16-0.19

transition temp

congealing temperature 34.5-37.5 °C (1.5% gel)

gel strength

≥1000 g/cm2 (1.0% gel)

SMILES string

O1[C@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O[C@@H]4O[C@@H]5[C@H]([C@@H](OC5)[C@@H]4O)O[C@@H]6O[C@@H]([C@@H]([C@@H]([C@H]6O)O)O)CO)O)O[C@H]2[C@H]3OC[C@@H]2O[C@H]([C@H]3O)O

InChI

1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1

InChI key

MJQHZNBUODTQTK-WKGBVCLCSA-N

General description

Agarose is a polymer extracted from agar or agar-bearing marine algae. This purified linear galactan hydrocolloid comprises alternating co-polymers D-galactose and 3,6-anhydro-L-galactose units connected by α-(1→3) and β-(1→4) glycosidic bonds. Agarose is highly biocompatible, possesses variable mechanical and diffusion properties.[1]

Application

Agarose has been used:
  • in gel electrophoresis
  • in the preparation of 2x agarose solution used along with 2x supplemented-MEM media as an overlay medium in plaque assay
  • in an immunodiffusion test for Paracoccidioidomycosis

Analysis Note

The following is a list of properties associated with our agaroses:
Sulfate content - used as an indicator of purity, since sulfate is the major ionic group present.
Gel strength - the force that must be applied to a gel to cause it to fracture.
Gel point - the temperature at which an aqueous agarose solution forms a gel as it cools. Agarose solutions exhibit hysteresis in the liquid-to-gel transition - that is, their gel point is not the same as their melting temperature.
Electroendosmosis (EEO) - a movement of liquid through the gel. Anionic groups in an agarose gel are affixed to the matrix and cannot move, but dissociable counter cations can migrate toward the cathode in the matrix, giving rise to EEO. Since electrophoretic movement of biopolymers is usually toward the anode, EEO can disrupt separations because of internal convection.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A9539A4718A2576
EEO

0.16-0.19

EEO

0.09-0.13

EEO

≤0.05

EEO

≤0.12

gel strength

≥1000 g/cm2 (1.0% gel)

gel strength

≥1200 g/cm2 (1% gel)

gel strength

-

gel strength

≥300 g/cm2 (1.5% gel)

form

powder

form

powder

form

powder

form

powder

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

grade

Molecular Biology

grade

Molecular Biology

grade

Molecular Biology

grade

Molecular Biology

impurities

≤10% moisture content

impurities

≤10% moisture content

impurities

-

impurities

≤7% water


Still not finding the right product?

Explore all of our products under Agarose


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters