Skip to Content
Merck
All Photos(3)

Documents

A5306

Sigma-Aldrich

Bacteriological agar

for molecular biology

Synonym(s):

Agar, Agar-agar, Gum agar

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H18O9)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
50405901
NACRES:
NA.31

grade

Molecular Biology

Quality Level

form

powder

InChI

1S/C14H24O9/c1-5-8(16)13-11(7(21-5)4-20-13)23-14-10(18)12(19-2)9(17)6(3-15)22-14/h5-18H,3-4H2,1-2H3/t5?,6-,7?,8-,9+,10-,11?,12+,13+,14?/m1/s1

InChI key

GYYDPBCUIJTIBM-DYOGSRDZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bacteriological agar is a colloidal substance and a cell wall component, obtained from Rhodoyceae (marine algae) and Gelidium sp. Agarose combined with agaropectin forms this polysaccharide. It is also called as agar-agar and is a polymer of galactoside. This agar is soluble in boiling water but not in cold water.

Application

Bacteriological agar has been used:
  • as a component of ATCC agar for Escherichia coli culture
  • as one of the experimental diet feed for Atlantic salmon fry, to study the protein and lysine requirements for maintenance and for tissue accretion
  • as component of yeast extract peptone dextrose medium, Luria broth medium and synthetic complete drop out medium for Escherichia coli culture

Biochem/physiol Actions

Bacteriological agar is commonly used as a culture medium for microorganism. It is useful for fermentation process. Agar-agar serves as a preservative in food processing. It also possesses various other applications such as an emulsifier, carrier, lubricant, stabilizer, laxative disintegrant in pharmaceutical and cosmetic industries. Agar-agar is also used in photographic emulsion.

Quality

A purified agar in which the original naturally occurring pigments, salts and miscellaneous matter have been reduced to a minimum.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natural Food Antimicrobial Systems, 417-417 (2000)
Measuring the activity of protein variants on a large scale using deep mutational scanning
Fowler DM, et al.
Nature Protocols, 9(9), 2267-2267 (2014)
Decontamination of materials contaminated with Francisella philomiragia or MS 2 bacteriophage using PES-Solid, a solid source of peracetic acid
Buhr TL, et al.
Journal of Applied Microbiology, 117(2), 397-404 (2014)
Protein and lysine requirements for maintenance and for tissue accretion in Atlantic salmon (Salmo salar) fry
Abboudi T, et al.
Aquaculture (Amsterdam, Netherlands), 261(1), 369-383 (2006)
Handbook of Fillers, Extenders, and Diluents, 223-223 (2007)

Protocols

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Magnetic-plasmonic nanoparticles: Synthesis and application of Ag/FeCo/Ag nanobeads for cellular organelle imaging and isolation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service