Skip to Content
Merck

A0701

Agarose, low gelling temperature

Type VII-A

Synonym(s):

2-Hydroxyethyl agarose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
41105317
NACRES:
NA.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


type

Type VII-A

Quality Segment

form

powder

technique(s)

electrophoresis: suitable

impurities

≤7% water

ash

≤0.4%

turbidity

≤4 NTU (1.5% gel)

EEO

≤0.12

mp

65 °C±1 °C

transition temp

gel point 26 °C ±2 °C (1.5% gel)

gel strength

≥250 g/cm2 (1% gel)

anion traces

sulfate (SO42-): ≤0.4%

SMILES string

O1[C@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O[C@@H]4O[C@@H]5[C@H]([C@@H](OC5)[C@@H]4O)O[C@@H]6O[C@@H]([C@@H]([C@@H]([C@H]6O)O)O)CO)O)O[C@H]2[C@H]3OC[C@@H]2O[C@H]([C@H]3O)O

InChI

1S/C24H38O19/c25-1-5-9(27)11(29)12(30)22(38-5)41-17-8-4-36-20(17)15(33)24(40-8)43-18-10(28)6(2-26)39-23(14(18)32)42-16-7-3-35-19(16)13(31)21(34)37-7/h5-34H,1-4H2/t5-,6-,7+,8+,9+,10+,11+,12-,13+,14-,15+,16-,17-,18+,19+,20+,21-,22+,23+,24+/m1/s1

InChI key

MJQHZNBUODTQTK-WKGBVCLCSA-N

General description

Agarose is a polymer extracted from agar or agar-bearing marine algae. This purified linear galactan hydrocolloid comprises alternating co-polymers D-galactose and 3,6-anhydro-L-galactopyranose units connected by α-(1→3) and β-(1→4) glycosidic bonds. Agarose is highly biocompatible due to its variable mechanical and diffusion properties.

Application

Excellent for in-gel enzymatic reactions and cloning assays and for recovery of heat-labile samples after electrophoresis
Agarose has been used:
  • to encapsulate Escherichia coli on a hydrogel in tissue culture
  • to entrap Aliivibrio fischeri on a disposable card involved in designing of toxicity biosensors
  • as a the dispersed phase of emulsion during preparation agar beads

Biochem/physiol Actions

Agarose can be used as a gelling agent, to separate nucleic acids electrophoretically because its gels have larger pore sizes than polyacrylamide gels at low concentrations. Unlike polyacrylamide, the consistency of the gels is more solid (but also less elastic) It is also employed to determine cross reaction in immunoelectrophoresis (IEP) and Ouchterlony (double diffusion) plates in which antibody-antigen precipitin lines are studied. Agarose is used to make gel plates or overlays for cells in tissue culture. In addition, it is also used to form a gel matrix (either beaded and/or crosslinked) which can be used in chromatographic separations.

Analysis Note

The following is a list of properties associated with our agaroses:
Sulfate content - used as an indicator of purity, since sulfate is the major ionic group present.
Gel strength - the force that must be applied to a gel to cause it to fracture.
Gel point - the temperature at which an aqueous agarose solution forms a gel as it cools. Agarose solutions exhibit hysteresis in the liquid-to-gel transition - that is, their gel point is not the same as their melting temperature.
Electroendosmosis (EEO) - a movement of liquid through the gel. Anionic groups in an agarose gel are affixed to the matrix and cannot move, but dissociable counter cations can migrate toward the cathode in the matrix, giving rise to EEO. Since electrophoretic movement of biopolymers is usually toward the anode, EEO can disrupt separations because of internal convection.

Other Notes

Discover LiChropur reagents ideal for HPLC or LC-MS analysis


Still not finding the right product?

Explore all of our products under Agarose, low gelling temperature


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library